Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595529_m1
          See other SLC25A25 GE Assays ›
          Gene Symbol:
          Slc25a25
          Gene Name:
          solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 25
          Gene Aliases:
          Mcsc, Pcscl
          Chromosome Location:
          Chr.3: 11442396 - 11476186 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595529_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc25a25
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145677.1 NP_663710.1 4 - 5 587 65
          XM_006233931.2 XP_006233993.1 5 - 6 744 65
          XM_006233932.2 XP_006233994.1 4 - 5 708 65
          XM_006233933.2 XP_006233995.1 5 - 6 1396 65
          XM_006233934.2 XP_006233996.1 4 - 5 1360 65
          XM_006233935.2 XP_006233997.1 5 - 6 1960 65
          XM_006233937.2 XP_006233999.1 5 - 6 980 65
          GenBank mRNA AY043169.1 - 4 - 5 587 65
          FQ211394.1 - 4 - 5 667 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002876.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc25a25

          Panther Classification:

          Molecular Function -

          mitochondrial carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          response to dietary excess
          response to activity
          response to food
          multicellular organism growth
          camera-type eye development
          cellular respiration
          ATP metabolic process
          adipose tissue development
          calcium ion transmembrane transport
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P506402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13299 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109606 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145790 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P506402 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P506402 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P506402 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P527238 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P506402 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61483 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10844351 - -
          Affymetrix Rat Gene 1.0 ST Array 10844342 - -
          Affymetrix Rat Gene 1.0 ST Array 10844343 - -
          Affymetrix Rat Gene 1.0 ST Array 10844344 - -
          Affymetrix Rat Gene 1.0 ST Array 10844350 - -
          Affymetrix Rat Gene 1.0 ST Array 10844345 - -
          Affymetrix Rat Gene 1.0 ST Array 10844341 - -
          Affymetrix Rat Gene 1.0 ST Array 10844347 - -
          Affymetrix Rat Gene 1.0 ST Array 10844349 - -
          Affymetrix Rat Gene 1.0 ST Array 10844346 - -
          Affymetrix Rat Gene 1.0 ST Array 10844340 - -
          Affymetrix Rat Gene 1.1 ST Array 10844339 - -
          Rat Genome 230 2.0 Array 1371754_at - -
          Rat Genome 230 2.0 Array 1371754_PM_at - -
          Affymetrix HT Rat Focus Array 1371754_PM_at - -
          Affymetrix HT Rat Focus Array 1371754_at - -
          Affymetrix Rat Clariom S Assay PSR0300022194.rn.1 TC0300002876.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300022195.rn.1 TC0300002876.rn.1 RTA TC
          Rat Expression Set 230 1371754_PM_at - -
          Rat Expression Set 230 1371754_at - -
          Affymetrix Rat Gene 1.0 ST Array 10844339 - -
          Rat Genome U34 Set rc_AA817759_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22163674 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367573 - -
          RatRef-12 Expression BeadChip ILMN_53974 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline