Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595596_m1
          See other TNFSF15 GE Assays ›
          Gene Symbol:
          Tnfsf15
          Gene Name:
          tumor necrosis factor (ligand) superfamily, member 15
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 79553935 - 79569974 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595596_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tnfsf15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145765.1 NP_665708.1 3 - 4 301 76
          GenBank mRNA AF520787.1 - 3 - 4 301 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002901.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tnfsf15

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of cysteine-type endopeptidase activity involved in apoptotic process
          immune response
          activation of NF-kappaB-inducing kinase activity
          cytokine metabolic process
          positive regulation of cytokine secretion
          death receptor binding
          cytokine activity
          tumor necrosis factor receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P129895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351278 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136351 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43508 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129895 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129895 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P129895 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P515211 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80370 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11692 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10877533 - -
          Affymetrix Rat Gene 1.0 ST Array 10877537 - -
          Affymetrix Rat Gene 1.0 ST Array 10877536 - -
          Affymetrix Rat Gene 1.0 ST Array 10877534 - -
          Affymetrix Rat Gene 1.1 ST Array 10877532 - -
          Affymetrix Rat Clariom S Assay PSR0500021717.rn.1 TC0500002901.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10877532 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21465662 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358015 - -
          RatRef-12 Expression BeadChip ILMN_64389 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline