Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595612_m1
          See other MAMDC4 GE Assays ›
          Gene Symbol:
          Mamdc4
          Gene Name:
          MAM domain containing 4
          Gene Aliases:
          Aegp
          Chromosome Location:
          Chr.3: 2786523 - 2795109 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595612_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mamdc4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145768.1 NP_665711.1 1 - 2 104 64
          GenBank mRNA L37380.1 - 1 - 2 104 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002688.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mamdc4

          Gene Ontology Categories:

          Function(s)

          protein transport

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P554965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682856 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106733 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103942 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P060051 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P060051 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P060051 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P554965 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12503 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10843507 - -
          Affymetrix Rat Gene 1.0 ST Array 10843497 - -
          Affymetrix Rat Gene 1.0 ST Array 10843494 - -
          Affymetrix Rat Gene 1.0 ST Array 10843514 - -
          Affymetrix Rat Gene 1.0 ST Array 10843508 - -
          Affymetrix Rat Gene 1.0 ST Array 10843516 - -
          Affymetrix Rat Gene 1.0 ST Array 10843505 - -
          Affymetrix Rat Gene 1.0 ST Array 10843518 - -
          Affymetrix Rat Gene 1.0 ST Array 10843499 - -
          Affymetrix Rat Gene 1.0 ST Array 10843498 - -
          Affymetrix Rat Gene 1.0 ST Array 10843520 - -
          Affymetrix Rat Gene 1.0 ST Array 10843515 - -
          Affymetrix Rat Gene 1.0 ST Array 10843510 - -
          Affymetrix Rat Gene 1.0 ST Array 10843511 - -
          Affymetrix Rat Gene 1.0 ST Array 10843509 - -
          Affymetrix Rat Gene 1.0 ST Array 10843506 - -
          Affymetrix Rat Gene 1.0 ST Array 10843503 - -
          Affymetrix Rat Gene 1.0 ST Array 10843522 - -
          Affymetrix Rat Gene 1.0 ST Array 10843504 - -
          Affymetrix Rat Gene 1.0 ST Array 10843517 - -
          Affymetrix Rat Gene 1.0 ST Array 10843502 - -
          Affymetrix Rat Gene 1.0 ST Array 10843496 - -
          Affymetrix Rat Gene 1.0 ST Array 10843512 - -
          Affymetrix Rat Gene 1.0 ST Array 10843501 - -
          Affymetrix Rat Gene 1.0 ST Array 10843500 - -
          Affymetrix Rat Gene 1.0 ST Array 10843521 - -
          Affymetrix Rat Gene 1.0 ST Array 10843513 - -
          Affymetrix Rat Gene 1.0 ST Array 10843495 - -
          Affymetrix Rat Gene 1.1 ST Array 10843493 - -
          Rat Genome 230 2.0 Array 1388017_PM_at - -
          Rat Genome 230 2.0 Array 1388017_at - -
          Affymetrix HT Rat Focus Array 1388017_PM_at - -
          Affymetrix HT Rat Focus Array 1388017_at - -
          Affymetrix Rat Clariom S Assay PSR0300020095.rn.1 TC0300002688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300020097.rn.1 TC0300002688.rn.1 RTA TC
          Rat Expression Set 230 1388017_PM_at - -
          Rat Expression Set 230 1388017_at - -
          Affymetrix Rat Gene 1.0 ST Array 10843493 - -
          Rat Genome U34 Set rc_AI171848_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22041593 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367662 - -
          RatRef-12 Expression BeadChip ILMN_63351 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline