Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595690_m1
          See other OLFM3 GE Assays ›
          Gene Symbol:
          Olfm3
          Gene Name:
          olfactomedin 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 217963376 - 218005244 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595690_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfm3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145777.1 NP_665720.1 4 - 5 746 89
          GenBank mRNA AF442822.1 - 4 - 5 746 89
          AF442823.1 - 637 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002033.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfm3

          Panther Classification:

          Molecular Function -

          structural protein

          Gene Ontology Categories:

          Function(s)

          eye photoreceptor cell development

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P675802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400119 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135630 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123640 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75387 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44151 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46493 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22032 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101809 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122137 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82329 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111022 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91619 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83494 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109915 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12545 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163463 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64069 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179924 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114292 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P675802 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P675802 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P675802 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P675802 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P456820 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75058 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35953 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94079 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83710 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90881 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4589 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64668 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44954 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71239 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48456 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21010 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24386 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94784 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10818578 - -
          Affymetrix Rat Gene 1.0 ST Array 10818576 - -
          Affymetrix Rat Gene 1.0 ST Array 10818574 - -
          Affymetrix Rat Gene 1.0 ST Array 10818579 - -
          Affymetrix Rat Gene 1.0 ST Array 10818577 - -
          Affymetrix Rat Gene 1.0 ST Array 10818575 - -
          Affymetrix Rat Gene 1.0 ST Array 10818580 - -
          Affymetrix Rat Gene 1.1 ST Array 10818573 - -
          Rat Genome 230 2.0 Array 1370455_PM_a_at - -
          Rat Genome 230 2.0 Array 1370455_a_at - -
          Affymetrix HT Rat Focus Array 1370455_PM_a_at - -
          Affymetrix HT Rat Focus Array 1370455_a_at - -
          Affymetrix Rat Clariom S Assay PSR0200014102.rn.1 TC0200002033.rn.1 RTA TC
          Rat Expression Set 230 1370455_a_at - -
          Rat Expression Set 230 1370455_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10818573 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21480570 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374487 - -
          RatRef-12 Expression BeadChip ILMN_57553 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline