Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595953_m1
          See other DCLRE1C GE Assays ›
          Gene Symbol:
          Dclre1c
          Gene Name:
          DNA cross-link repair 1C
          Gene Aliases:
          Snm1l
          Chromosome Location:
          Chr.17: 78783618 - 78812059 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595953_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dclre1c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147145.2 NP_671486.2 8 - 9 698 105
          GenBank mRNA AF395746.1 - 8 - 9 730 105
          BC083546.1 - 8 - 9 698 7134940 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700002239.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dclre1c

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance
          DNA catabolic process, endonucleolytic
          double-strand break repair
          DNA recombination
          biological_process
          response to ionizing radiation
          B cell differentiation
          V(D)J recombination
          chromosome organization
          nucleic acid phosphodiester bond hydrolysis
          single-stranded DNA endodeoxyribonuclease activity
          molecular_function
          5'-3' exonuclease activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P783205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686859 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153561 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47498 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82020 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P783205 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P783205 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P783205 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P783205 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53772 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48914 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10799643 - -
          Affymetrix Rat Gene 1.0 ST Array 10799639 - -
          Affymetrix Rat Gene 1.0 ST Array 10799644 - -
          Affymetrix Rat Gene 1.0 ST Array 10799635 - -
          Affymetrix Rat Gene 1.0 ST Array 10799638 - -
          Affymetrix Rat Gene 1.0 ST Array 10799633 - -
          Affymetrix Rat Gene 1.0 ST Array 10799631 - -
          Affymetrix Rat Gene 1.0 ST Array 10799642 - -
          Affymetrix Rat Gene 1.0 ST Array 10799637 - -
          Affymetrix Rat Gene 1.0 ST Array 10799640 - -
          Affymetrix Rat Gene 1.0 ST Array 10799630 - -
          Affymetrix Rat Gene 1.0 ST Array 10799634 - -
          Affymetrix Rat Gene 1.0 ST Array 10799641 - -
          Affymetrix Rat Gene 1.0 ST Array 10799636 - -
          Affymetrix Rat Gene 1.0 ST Array 10799632 - -
          Affymetrix Rat Gene 1.1 ST Array 10799627 - -
          Rat Genome 230 2.0 Array 1383597_at - -
          Rat Genome 230 2.0 Array 1385776_at - -
          Rat Genome 230 2.0 Array 1383597_PM_at - -
          Rat Genome 230 2.0 Array 1385776_PM_at - -
          Affymetrix HT Rat Focus Array 1383597_PM_at - -
          Affymetrix HT Rat Focus Array 1383597_at - -
          Affymetrix Rat Clariom S Assay PSR1700014914.rn.1 TC1700002239.rn.1 RTA TC
          Rat Expression Set 230 1383597_at - -
          Rat Expression Set 230 1383597_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10799627 - -
          Rat Genome U34 Set rc_AI072301_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21999200 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366473 - -
          RatRef-12 Expression BeadChip ILMN_66197 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline