Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00596111_m1
          See other SNX27 GE Assays ›
          Gene Symbol:
          Snx27
          Gene Name:
          sorting nexin family member 27
          Gene Aliases:
          Mrt1
          Chromosome Location:
          Chr.2: 195738613 - 195821608 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00596111_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Snx27
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110151.1 NP_001103621.1 1 - 2 427 128
          NM_152847.2 NP_690060.2 1 - 2 427 128
          GenBank mRNA AB010245.1 - 1 - 2 427 128
          AB051816.1 - 1 - 2 427 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004184.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Snx27

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          signal transduction
          nervous system development
          endosome to lysosome transport
          endosomal transport
          response to drug
          retrograde transport, endosome to plasma membrane
          protein binding
          phosphatidylinositol-3-phosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P125938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565591 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73810 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130492 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106721 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114272 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85754 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67862 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160185 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118877 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159166 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P125938 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P125938 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P125938 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P311468 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P728389 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15081 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33608 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9040 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26763 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103710 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80906 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4414 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10824839 - -
          Affymetrix Rat Gene 1.0 ST Array 10824836 - -
          Affymetrix Rat Gene 1.0 ST Array 10824833 - -
          Affymetrix Rat Gene 1.0 ST Array 10824840 - -
          Affymetrix Rat Gene 1.0 ST Array 10824834 - -
          Affymetrix Rat Gene 1.0 ST Array 10824832 - -
          Affymetrix Rat Gene 1.0 ST Array 10824835 - -
          Affymetrix Rat Gene 1.0 ST Array 10824837 - -
          Affymetrix Rat Gene 1.0 ST Array 10824830 - -
          Affymetrix Rat Gene 1.0 ST Array 10824838 - -
          Affymetrix Rat Gene 1.0 ST Array 10824828 - -
          Affymetrix Rat Gene 1.0 ST Array 10824841 - -
          Affymetrix Rat Gene 1.0 ST Array 10824829 - -
          Affymetrix Rat Gene 1.0 ST Array 10824831 - -
          Affymetrix Rat Gene 1.1 ST Array 10824827 - -
          Rat Genome 230 2.0 Array 1377526_at - -
          Rat Genome 230 2.0 Array 1381394_at - -
          Rat Genome 230 2.0 Array 1379804_PM_at - -
          Rat Genome 230 2.0 Array 1381394_PM_at - -
          Rat Genome 230 2.0 Array 1372203_at - -
          Rat Genome 230 2.0 Array 1379804_at - -
          Rat Genome 230 2.0 Array 1377526_PM_at - -
          Rat Genome 230 2.0 Array 1372203_PM_at - -
          Affymetrix HT Rat Focus Array 1372203_at - -
          Affymetrix HT Rat Focus Array 1379804_PM_at - -
          Affymetrix HT Rat Focus Array 1372203_PM_at - -
          Affymetrix HT Rat Focus Array 1381394_at - -
          Affymetrix HT Rat Focus Array 1379804_at - -
          Affymetrix HT Rat Focus Array 1377526_at - -
          Affymetrix HT Rat Focus Array 1381394_PM_at - -
          Affymetrix HT Rat Focus Array 1377526_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200028509.rn.1 TC0200004184.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200028514.rn.1 TC0200004184.rn.1 RTA TC
          Rat Expression Set 230 1377526_at - -
          Rat Expression Set 230 1377526_PM_at - -
          Rat Expression Set 230 1372203_PM_at - -
          Rat Expression Set 230 1372203_at - -
          Rat Expression Set 230 1381394_at - -
          Rat Expression Set 230 1381394_PM_at - -
          Rat Expression Set 230 1379804_at - -
          Rat Expression Set 230 1379804_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10824827 - -
          Rat Genome U34 Set rc_AA900057_at - -
          Rat Genome U34 Set rc_AI101286_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22012938 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353338 - -
          RatRef-12 Expression BeadChip ILMN_56991 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline