Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00596158_m1
          See other RBM10 GE Assays ›
          Gene Symbol:
          Rbm10
          Gene Name:
          RNA binding motif protein 10
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 1754869 - 1786973 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00596158_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbm10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_152861.4 NP_690600.1 5 - 6 687 57
          GenBank mRNA D83948.1 - 5 - 6 687 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001383.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rbm10

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          negative regulation of cell proliferation
          regulation of smooth muscle cell apoptotic process
          positive regulation of smooth muscle cell apoptotic process
          negative regulation of mRNA splicing, via spliceosome
          3'-UTR-mediated mRNA stabilization
          nucleotide binding
          zinc ion binding
          protein complex binding
          identical protein binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P540065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591859 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143427 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45883 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98657 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2270 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174622 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P540065 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P540065 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P540065 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P540065 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P540067 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700370154 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104795 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68435 - -
          Agilent Rat cDNA Microarray (G4105A) 700370154 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10936631 - -
          Affymetrix Rat Gene 1.0 ST Array 10936629 - -
          Affymetrix Rat Gene 1.0 ST Array 10936624 - -
          Affymetrix Rat Gene 1.0 ST Array 10936617 - -
          Affymetrix Rat Gene 1.0 ST Array 10936615 - -
          Affymetrix Rat Gene 1.0 ST Array 10936622 - -
          Affymetrix Rat Gene 1.0 ST Array 10936613 - -
          Affymetrix Rat Gene 1.0 ST Array 10936621 - -
          Affymetrix Rat Gene 1.0 ST Array 10936616 - -
          Affymetrix Rat Gene 1.0 ST Array 10936625 - -
          Affymetrix Rat Gene 1.0 ST Array 10936627 - -
          Affymetrix Rat Gene 1.0 ST Array 10936623 - -
          Affymetrix Rat Gene 1.0 ST Array 10936633 - -
          Affymetrix Rat Gene 1.0 ST Array 10936614 - -
          Affymetrix Rat Gene 1.0 ST Array 10936619 - -
          Affymetrix Rat Gene 1.0 ST Array 10936618 - -
          Affymetrix Rat Gene 1.0 ST Array 10936628 - -
          Affymetrix Rat Gene 1.0 ST Array 10936634 - -
          Affymetrix Rat Gene 1.0 ST Array 10936612 - -
          Affymetrix Rat Gene 1.0 ST Array 10936636 - -
          Affymetrix Rat Gene 1.0 ST Array 10936620 - -
          Affymetrix Rat Gene 1.0 ST Array 10936611 - -
          Affymetrix Rat Gene 1.0 ST Array 10936630 - -
          Affymetrix Rat Gene 1.1 ST Array 10936610 - -
          Rat Genome 230 2.0 Array 1385478_at - -
          Rat Genome 230 2.0 Array 1370356_PM_at - -
          Rat Genome 230 2.0 Array 1370356_at - -
          Rat Genome 230 2.0 Array 1385478_PM_at - -
          Affymetrix HT Rat Focus Array 1385478_at - -
          Affymetrix HT Rat Focus Array 1370356_PM_at - -
          Affymetrix HT Rat Focus Array 1385478_PM_at - -
          Affymetrix HT Rat Focus Array 1370356_at - -
          Affymetrix Rat Clariom S Assay PSR0X00008762.rn.1 TC0X00001383.rn.1 RTA TC
          Rat Expression Set 230 1370356_PM_at - -
          Rat Expression Set 230 1370356_at - -
          Rat Expression Set 230 1385478_PM_at - -
          Rat Expression Set 230 1385478_at - -
          Affymetrix Rat Gene 1.0 ST Array 10936610 - -
          Rat Genome U34 Set D83948mRNA_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22192171 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349535 - -
          RatRef-12 Expression BeadChip ILMN_54143 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline