Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00596209_m1
          See other COPS2 GE Assays ›
          Gene Symbol:
          Cops2
          Gene Name:
          COP9 signalosome subunit 2
          Gene Aliases:
          CSN2, Trip15
          Chromosome Location:
          Chr.3: 118109394 - 118137125 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00596209_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cops2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153297.1 NP_695209.1 3 - 4 396 108
          XM_006234901.2 XP_006234963.1 3 - 4 325 108
          XM_008762146.1 XP_008760368.1 3 - 4 325 108
          GenBank mRNA AB081072.1 - 3 - 4 396 108
          BC061864.1 - 3 - 4 296 5597874 108
          FQ210124.1 - 3 - 4 272 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004081.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cops2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cell proliferation
          cullin deneddylation
          neuron differentiation
          skeletal muscle cell differentiation
          negative regulation of transcription, DNA-templated
          transcription corepressor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P344306 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P017861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454033 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13881 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132089 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017861 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P344306 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017861 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P344306 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P017861 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P344306 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P344306 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12485 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10849470 - -
          Affymetrix Rat Gene 1.0 ST Array 10849457 - -
          Affymetrix Rat Gene 1.0 ST Array 10849468 - -
          Affymetrix Rat Gene 1.0 ST Array 10849461 - -
          Affymetrix Rat Gene 1.0 ST Array 10849469 - -
          Affymetrix Rat Gene 1.0 ST Array 10849458 - -
          Affymetrix Rat Gene 1.0 ST Array 10849471 - -
          Affymetrix Rat Gene 1.0 ST Array 10849464 - -
          Affymetrix Rat Gene 1.0 ST Array 10849463 - -
          Affymetrix Rat Gene 1.0 ST Array 10849462 - -
          Affymetrix Rat Gene 1.0 ST Array 10849460 - -
          Affymetrix Rat Gene 1.0 ST Array 10849456 - -
          Affymetrix Rat Gene 1.0 ST Array 10849465 - -
          Affymetrix Rat Gene 1.0 ST Array 10849459 - -
          Affymetrix Rat Gene 1.0 ST Array 10849467 - -
          Affymetrix Rat Gene 1.0 ST Array 10849466 - -
          Affymetrix Rat Gene 1.1 ST Array 10849454 - -
          Rat Genome 230 2.0 Array 1383819_PM_at - -
          Rat Genome 230 2.0 Array 1384366_PM_at - -
          Rat Genome 230 2.0 Array 1384366_at - -
          Rat Genome 230 2.0 Array 1383819_at - -
          Rat Genome 230 2.0 Array 1373029_PM_at - -
          Rat Genome 230 2.0 Array 1373029_at - -
          Affymetrix HT Rat Focus Array 1373029_at - -
          Affymetrix HT Rat Focus Array 1373029_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300031573.rn.1 TC0300004081.rn.1 RTA TC
          Rat Expression Set 230 1373029_at - -
          Rat Expression Set 230 1373029_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10849454 - -
          Rat Genome U34 Set rc_AI169377_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22281930 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359102 - -
          RatRef-12 Expression BeadChip ILMN_65553 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline