Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00596354_m1
          See other DHRS4 GE Assays ›
          Gene Symbol:
          Dhrs4
          Gene Name:
          dehydrogenase/reductase (SDR family) member 4
          Gene Aliases:
          PSCD
          Chromosome Location:
          Chr.15: 34155255 - 34167073 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00596354_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dhrs4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153315.2 NP_695227.2 2 - 3 438 77
          GenBank mRNA AB062758.1 - 2 - 3 254 77
          BC070961.1 - 2 - 3 438 7099044 77

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dhrs4

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          alcohol metabolic process
          steroid metabolic process
          cellular ketone metabolic process
          retinal metabolic process
          protein tetramerization
          oxidation-reduction process
          3-keto sterol reductase activity
          retinal dehydrogenase activity
          carbonyl reductase (NADPH) activity
          receptor binding
          oxidoreductase activity, acting on NAD(P)H, quinone or similar compound as acceptor
          alcohol dehydrogenase [NAD(P)+] activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P465868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246636 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62344 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17567 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P465868 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P465868 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P465868 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P465868 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89725 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48065 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10780290 - -
          Affymetrix Rat Gene 1.0 ST Array 10780300 - -
          Affymetrix Rat Gene 1.0 ST Array 10780299 - -
          Affymetrix Rat Gene 1.0 ST Array 10780297 - -
          Affymetrix Rat Gene 1.0 ST Array 10780293 - -
          Affymetrix Rat Gene 1.0 ST Array 10780298 - -
          Affymetrix Rat Gene 1.0 ST Array 10780294 - -
          Affymetrix Rat Gene 1.0 ST Array 10780291 - -
          Affymetrix Rat Gene 1.0 ST Array 10780292 - -
          Affymetrix Rat Gene 1.0 ST Array 10780295 - -
          Affymetrix Rat Gene 1.1 ST Array 10780289 - -
          Rat Genome 230 2.0 Array 1385488_PM_at - -
          Rat Genome 230 2.0 Array 1370814_PM_at - -
          Rat Genome 230 2.0 Array 1385488_at - -
          Rat Genome 230 2.0 Array 1370814_at - -
          Affymetrix HT Rat Focus Array 1385488_PM_at - -
          Affymetrix HT Rat Focus Array 1370814_PM_at - -
          Affymetrix HT Rat Focus Array 1370814_at - -
          Affymetrix HT Rat Focus Array 1385488_at - -
          Rat Expression Set 230 1370814_at - -
          Rat Expression Set 230 1370814_PM_at - -
          Rat Expression Set 230 1385488_at - -
          Rat Expression Set 230 1385488_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10780289 - -
          Rat Genome U34 Set rc_AI103224_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22257078 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361326 - -
          RatRef-12 Expression BeadChip ILMN_57803 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline