Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00596371_m1
          See other CYP4F6 GE Assays ›
          Gene Symbol:
          Cyp4f6
          Gene Name:
          cytochrome P450, family 4, subfamily f, polypeptide 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 15198950 - 15225552 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00596371_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp4f6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153318.1 NP_695230.1 3 - 4 466 67
          XM_006241049.1 XP_006241111.1 3 - 4 540 67
          XM_008765156.1 XP_008763378.1 3 - 4 619 67
          GenBank mRNA U39208.1 - 3 - 4 466 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002255.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 P450 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp4f6

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          leukotriene metabolic process
          inflammatory response
          oxidation-reduction process
          monooxygenase activity
          iron ion binding
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          heme binding
          aromatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P058850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680317 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174500 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80517 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121209 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058850 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058850 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P058850 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P814479 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700396386 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92586 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43072 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34655 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21655 - -
          Agilent Rat cDNA Microarray (G4105A) 700396386 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10901076 - -
          Affymetrix Rat Gene 1.0 ST Array 10901072 - -
          Affymetrix Rat Gene 1.0 ST Array 10901081 - -
          Affymetrix Rat Gene 1.0 ST Array 10901079 - -
          Affymetrix Rat Gene 1.0 ST Array 10901077 - -
          Affymetrix Rat Gene 1.0 ST Array 10901075 - -
          Affymetrix Rat Gene 1.0 ST Array 10901082 - -
          Affymetrix Rat Gene 1.0 ST Array 10901078 - -
          Affymetrix Rat Gene 1.0 ST Array 10901080 - -
          Affymetrix Rat Gene 1.0 ST Array 10901070 - -
          Affymetrix Rat Gene 1.0 ST Array 10901074 - -
          Affymetrix Rat Gene 1.0 ST Array 10901071 - -
          Affymetrix Rat Gene 1.0 ST Array 10901073 - -
          Affymetrix Rat Gene 1.1 ST Array 10901069 - -
          Rat Genome 230 2.0 Array 1387916_at - -
          Rat Genome 230 2.0 Array 1387916_PM_at - -
          Affymetrix HT Rat Focus Array 1387916_at - -
          Affymetrix HT Rat Focus Array 1387916_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700018179.rn.1 TC0700002255.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700018180.rn.1 TC0700002255.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700018177.rn.1 TC0700002255.rn.1 RTA TC
          Rat Expression Set 230 1387916_PM_at - -
          Rat Expression Set 230 1387916_at - -
          Affymetrix Rat Gene 1.0 ST Array 10901069 - -
          Rat Genome U34 Set U39208_at - -
          Rat Genome U34 Set U39208_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20872342 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366152 - -
          RatRef-12 Expression BeadChip ILMN_50203 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline