Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00596415_m1
          See other RNF40 GE Assays ›
          Gene Symbol:
          Rnf40
          Gene Name:
          ring finger protein 40, E3 ubiquitin protein ligase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 199037481 - 199052823 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00596415_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf40
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153471.1 NP_703201.1 19 - 20 2927 93
          XM_006230219.2 XP_006230281.1 19 - 20 3005 93
          GenBank mRNA AF352815.1 - 19 - 20 2927 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002649.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf40

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          regulation of mitotic cell cycle
          histone monoubiquitination
          histone H2B ubiquitination
          negative regulation of mRNA polyadenylation
          positive regulation of histone H2B ubiquitination
          mRNA 3'-UTR binding
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          ligase activity
          syntaxin-1 binding
          ubiquitin protein ligase binding
          protein complex binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P045578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184481 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141690 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6664 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P045578 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P045578 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P045578 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P538483 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54992 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39955 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10711145 - -
          Affymetrix Rat Gene 1.0 ST Array 10711158 - -
          Affymetrix Rat Gene 1.0 ST Array 10711149 - -
          Affymetrix Rat Gene 1.0 ST Array 10711155 - -
          Affymetrix Rat Gene 1.0 ST Array 10711154 - -
          Affymetrix Rat Gene 1.0 ST Array 10711140 - -
          Affymetrix Rat Gene 1.0 ST Array 10711141 - -
          Affymetrix Rat Gene 1.0 ST Array 10711153 - -
          Affymetrix Rat Gene 1.0 ST Array 10711142 - -
          Affymetrix Rat Gene 1.0 ST Array 10711150 - -
          Affymetrix Rat Gene 1.0 ST Array 10711151 - -
          Affymetrix Rat Gene 1.0 ST Array 10711157 - -
          Affymetrix Rat Gene 1.0 ST Array 10711143 - -
          Affymetrix Rat Gene 1.0 ST Array 10711147 - -
          Affymetrix Rat Gene 1.0 ST Array 10711152 - -
          Affymetrix Rat Gene 1.0 ST Array 10711156 - -
          Affymetrix Rat Gene 1.0 ST Array 10711146 - -
          Affymetrix Rat Gene 1.0 ST Array 10711148 - -
          Affymetrix Rat Gene 1.0 ST Array 10711144 - -
          Affymetrix Rat Gene 1.1 ST Array 10711139 - -
          Rat Genome 230 2.0 Array 1393731_at - -
          Rat Genome 230 2.0 Array 1393731_PM_at - -
          Affymetrix HT Rat Focus Array 1393731_PM_at - -
          Affymetrix HT Rat Focus Array 1393731_at - -
          Affymetrix Rat Clariom S Assay PSR0100020614.rn.1 TC0100002649.rn.1 RTA TC
          Rat Expression Set 230 1393731_at - -
          Rat Expression Set 230 1393731_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10711139 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20876299 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360868 - -
          RatRef-12 Expression BeadChip ILMN_52172 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline