Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00596426_m1
          See other MNAT1 GE Assays ›
          Gene Symbol:
          Mnat1
          Gene Name:
          MNAT CDK-activating kinase assembly factor 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 96007805 - 96153331 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00596426_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mnat1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153472.2 NP_703202.1 4 - 5 476 125
          GenBank mRNA AY135567.1 - 4 - 5 431 125
          BC078712.1 - 4 - 5 476 7128354 125

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600000972.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mnat1

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cyclin-dependent protein serine/threonine kinase activity
          G1/S transition of mitotic cell cycle
          DNA repair
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          protein phosphorylation
          adult heart development
          ventricular system development
          negative regulation of apoptotic process
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of smooth muscle cell proliferation
          response to calcium ion
          molecular_function
          DNA-dependent ATPase activity
          zinc ion binding
          RNA polymerase II carboxy-terminal domain kinase activity
          protein N-terminus binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P047162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252891 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22269 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7895 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47831 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13556 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157872 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155994 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57651 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148464 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144438 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102728 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129031 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113176 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55128 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13473 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79686 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63871 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82534 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135319 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137569 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112898 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156264 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170221 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047162 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047162 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P047162 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P727530 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60856 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92825 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18410 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68941 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37980 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30629 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52857 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11069 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87441 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22516 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15311 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10885210 - -
          Affymetrix Rat Gene 1.0 ST Array 10885216 - -
          Affymetrix Rat Gene 1.0 ST Array 10885213 - -
          Affymetrix Rat Gene 1.0 ST Array 10885214 - -
          Affymetrix Rat Gene 1.0 ST Array 10885215 - -
          Affymetrix Rat Gene 1.0 ST Array 10885209 - -
          Affymetrix Rat Gene 1.0 ST Array 10885212 - -
          Affymetrix Rat Gene 1.0 ST Array 10885211 - -
          Affymetrix Rat Gene 1.0 ST Array 10885208 - -
          Affymetrix Rat Gene 1.1 ST Array 10885207 - -
          Rat Genome 230 2.0 Array 1375956_at - -
          Rat Genome 230 2.0 Array 1375956_PM_at - -
          Affymetrix HT Rat Focus Array 1375956_PM_at - -
          Affymetrix HT Rat Focus Array 1375956_at - -
          Affymetrix Rat Clariom S Assay PSR0600006587.rn.1 TC0600000972.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600006595.rn.1 TC0600000972.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600006597.rn.1 TC0600000972.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600006593.rn.1 TC0600000972.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600006588.rn.1 TC0600000972.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600006589.rn.1 TC0600000972.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600006591.rn.1 TC0600000972.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600006592.rn.1 TC0600000972.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600006596.rn.1 TC0600000972.rn.1 RTA TC
          Rat Expression Set 230 1375956_PM_at - -
          Rat Expression Set 230 1375956_at - -
          Affymetrix Rat Gene 1.0 ST Array 10885207 - -
          Rat Genome U34 Set rc_AI104569_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20752441 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369015 - -
          RatRef-12 Expression BeadChip ILMN_67889 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline