Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00596450_m1
          See other MYO7A GE Assays ›
          Gene Symbol:
          Myo7a
          Gene Name:
          myosin VIIA
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 163001313 - 163071542 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00596450_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Myo7a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153473.1 NP_703203.1 32 - 33 4596 70
          XM_006229742.2 XP_006229804.1 31 - 32 5402 70
          XM_006229743.2 XP_006229805.1 31 - 32 5401 70
          XM_006229744.2 XP_006229806.1 32 - 33 4630 70
          GenBank mRNA AB091825.1 - 32 - 33 4596 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006084.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Myo7a

          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          phagolysosome assembly
          intracellular protein transport
          phagocytosis
          lysosome organization
          sensory perception
          visual perception
          sensory perception of sound
          metabolic process
          cell projection organization
          actin filament-based movement
          inner ear morphogenesis
          mechanoreceptor differentiation
          auditory receptor cell differentiation
          post-embryonic organ morphogenesis
          inner ear development
          sensory perception of light stimulus
          equilibrioception
          pigment granule localization
          pigment granule transport
          auditory receptor cell stereocilium organization
          inner ear receptor cell differentiation
          microfilament motor activity
          motor activity
          actin binding
          calmodulin binding
          ATP binding
          protein domain specific binding
          spectrin binding
          actin-dependent ATPase activity
          protein complex binding
          protein homodimerization activity
          ADP binding
          protein N-terminus binding
          actin filament binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P113695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957636 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100440 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23630 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177671 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125027 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34535 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156201 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134059 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P113695 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P113695 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P113695 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P418942 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64675 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38536 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15277 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61066 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104518 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10723757 - -
          Affymetrix Rat Gene 1.0 ST Array 10723746 - -
          Affymetrix Rat Gene 1.0 ST Array 10723741 - -
          Affymetrix Rat Gene 1.0 ST Array 10723789 - -
          Affymetrix Rat Gene 1.0 ST Array 10723763 - -
          Affymetrix Rat Gene 1.0 ST Array 10723791 - -
          Affymetrix Rat Gene 1.0 ST Array 10723782 - -
          Affymetrix Rat Gene 1.0 ST Array 10723742 - -
          Affymetrix Rat Gene 1.0 ST Array 10723786 - -
          Affymetrix Rat Gene 1.0 ST Array 10723766 - -
          Affymetrix Rat Gene 1.0 ST Array 10723748 - -
          Affymetrix Rat Gene 1.0 ST Array 10723778 - -
          Affymetrix Rat Gene 1.0 ST Array 10723745 - -
          Affymetrix Rat Gene 1.0 ST Array 10723750 - -
          Affymetrix Rat Gene 1.0 ST Array 10723774 - -
          Affymetrix Rat Gene 1.0 ST Array 10723777 - -
          Affymetrix Rat Gene 1.0 ST Array 10723776 - -
          Affymetrix Rat Gene 1.0 ST Array 10723773 - -
          Affymetrix Rat Gene 1.0 ST Array 10723768 - -
          Affymetrix Rat Gene 1.0 ST Array 10723760 - -
          Affymetrix Rat Gene 1.0 ST Array 10723775 - -
          Affymetrix Rat Gene 1.0 ST Array 10723770 - -
          Affymetrix Rat Gene 1.0 ST Array 10723765 - -
          Affymetrix Rat Gene 1.0 ST Array 10723758 - -
          Affymetrix Rat Gene 1.0 ST Array 10723744 - -
          Affymetrix Rat Gene 1.0 ST Array 10723754 - -
          Affymetrix Rat Gene 1.0 ST Array 10723781 - -
          Affymetrix Rat Gene 1.0 ST Array 10723762 - -
          Affymetrix Rat Gene 1.0 ST Array 10723752 - -
          Affymetrix Rat Gene 1.0 ST Array 10723780 - -
          Affymetrix Rat Gene 1.0 ST Array 10723787 - -
          Affymetrix Rat Gene 1.0 ST Array 10723755 - -
          Affymetrix Rat Gene 1.0 ST Array 10723790 - -
          Affymetrix Rat Gene 1.0 ST Array 10723769 - -
          Affymetrix Rat Gene 1.0 ST Array 10723747 - -
          Affymetrix Rat Gene 1.0 ST Array 10723743 - -
          Affymetrix Rat Gene 1.0 ST Array 10723771 - -
          Affymetrix Rat Gene 1.0 ST Array 10723759 - -
          Affymetrix Rat Gene 1.0 ST Array 10723772 - -
          Affymetrix Rat Gene 1.0 ST Array 10723749 - -
          Affymetrix Rat Gene 1.0 ST Array 10723783 - -
          Affymetrix Rat Gene 1.0 ST Array 10723788 - -
          Affymetrix Rat Gene 1.0 ST Array 10723761 - -
          Affymetrix Rat Gene 1.0 ST Array 10723779 - -
          Affymetrix Rat Gene 1.0 ST Array 10723784 - -
          Affymetrix Rat Gene 1.0 ST Array 10723751 - -
          Affymetrix Rat Gene 1.0 ST Array 10723753 - -
          Affymetrix Rat Gene 1.0 ST Array 10723785 - -
          Affymetrix Rat Gene 1.0 ST Array 10723767 - -
          Affymetrix Rat Gene 1.1 ST Array 10723740 - -
          Affymetrix Rat Clariom S Assay PSR0100048274.rn.1 TC0100006084.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10723740 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20846498 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369399 - -
          RatRef-12 Expression BeadChip ILMN_69659 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline