Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00596866_m1
          See other DECR2 GE Assays ›
          Gene Symbol:
          Decr2
          Gene Name:
          2,4-dienoyl CoA reductase 2, peroxisomal
          Gene Aliases:
          Dcrakl, putativeperoxisomal24-dienoyl-CoAreductase
          Chromosome Location:
          Chr.10: 15451850 - 15460215 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00596866_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Decr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_171996.3 NP_741993.1 4 - 5 429 76
          XM_006246024.2 XP_006246086.1 5 - 6 629 76
          GenBank mRNA AF044574.1 - 4 - 5 351 76
          BC070959.1 - 4 - 5 430 7097883 76
          FQ218511.1 - 4 - 5 404 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002453.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Decr2

          Panther Classification:

          Molecular Function -

          reductase

          Gene Ontology Categories:

          Function(s) Process(es)

          unsaturated fatty acid biosynthetic process
          oxidation-reduction process
          receptor binding
          2,4-dienoyl-CoA reductase (NADPH) activity
          trans-2-enoyl-CoA reductase (NADPH) activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P378631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141445 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45570 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P378631 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P378631 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P378631 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P378631 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701030993 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701246587 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56600 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40487 - -
          Agilent Rat cDNA Microarray (G4105A) 701246587 - -
          Agilent Rat cDNA Microarray (G4105A) 701030993 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10741713 - -
          Affymetrix Rat Gene 1.0 ST Array 10741707 - -
          Affymetrix Rat Gene 1.0 ST Array 10741715 - -
          Affymetrix Rat Gene 1.0 ST Array 10741706 - -
          Affymetrix Rat Gene 1.0 ST Array 10741714 - -
          Affymetrix Rat Gene 1.0 ST Array 10741708 - -
          Affymetrix Rat Gene 1.0 ST Array 10741712 - -
          Affymetrix Rat Gene 1.0 ST Array 10741705 - -
          Affymetrix Rat Gene 1.0 ST Array 10741716 - -
          Affymetrix Rat Gene 1.0 ST Array 10741709 - -
          Affymetrix Rat Gene 1.0 ST Array 10741711 - -
          Affymetrix Rat Gene 1.0 ST Array 10741710 - -
          Affymetrix Rat Gene 1.1 ST Array 10741701 - -
          Rat Genome 230 2.0 Array 1370818_PM_at - -
          Rat Genome 230 2.0 Array 1389129_at - -
          Rat Genome 230 2.0 Array 1389129_PM_at - -
          Rat Genome 230 2.0 Array 1370818_at - -
          Affymetrix HT Rat Focus Array 1370818_PM_at - -
          Affymetrix HT Rat Focus Array 1370818_at - -
          Affymetrix Rat Clariom S Assay PSR1000022303.rn.1 TC1000002453.rn.1 RTA TC
          Rat Expression Set 230 1389129_at - -
          Rat Expression Set 230 1389129_PM_at - -
          Rat Expression Set 230 1370818_PM_at - -
          Rat Expression Set 230 1370818_at - -
          Affymetrix Rat Gene 1.0 ST Array 10741701 - -
          Rat Genome U34 Set AF044574_at - -
          Rat Genome U34 Set AF044574_g_at - -
          Rat Genome U34 Set rc_AA892367_i_at - -
          Rat Genome U34 Set rc_AA945578_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21480204 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352032 - -
          RatRef-12 Expression BeadChip ILMN_69996 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline