Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00596890_m1
          See other PPP1R14D GE Assays ›
          Gene Symbol:
          Ppp1r14d
          Gene Name:
          protein phosphatase 1, regulatory (inhibitor) subunit 14D
          Gene Aliases:
          Gbpi
          Chromosome Location:
          Chr.3: 111022920 - 111037166 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00596890_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppp1r14d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172011.1 NP_742008.1 1 - 2 298 66
          GenBank mRNA AY122322.1 - 1 - 2 298 66

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppp1r14d

          Panther Classification:

          Molecular Function -

          phosphatase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of protein dephosphorylation
          regulation of phosphorylation
          negative regulation of catalytic activity
          protein phosphatase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P108800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866710 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150549 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27233 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P327882 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P327882 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P327882 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P327882 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47701 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37901 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10848529 - -
          Affymetrix Rat Gene 1.0 ST Array 10848528 - -
          Affymetrix Rat Gene 1.0 ST Array 10848530 - -
          Affymetrix Rat Gene 1.0 ST Array 10848532 - -
          Affymetrix Rat Gene 1.1 ST Array 10848527 - -
          Affymetrix Rat Gene 1.0 ST Array 10848527 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22103058 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358046 - -
          RatRef-12 Expression BeadChip ILMN_55970 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline