Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00596920_m1
          See other LZTS3 GE Assays ›
          Gene Symbol:
          Lzts3
          Gene Name:
          leucine zipper, putative tumor suppressor family member 3
          Gene Aliases:
          Prosapip1
          Chromosome Location:
          Chr.3: 123174444 - 123185643 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00596920_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lzts3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172022.2 NP_742019.2 2 - 3 559 57
          XM_006235009.2 XP_006235071.1 4 - 5 1788 57
          XM_006235010.2 XP_006235072.1 4 - 5 2540 57
          XM_006235011.2 XP_006235073.1 3 - 4 1740 57
          XM_006235014.2 XP_006235076.1 3 - 4 610 57
          XM_008762147.1 XP_008760369.1 4 - 5 1577 57
          XM_008762148.1 XP_008760370.1 3 - 4 1528 57
          XM_008762149.1 XP_008760371.1 4 - 5 1578 57
          XM_008762150.1 XP_008760372.1 3 - 4 1529 57
          XM_008762151.1 XP_008760373.1 2 - 3 601 57
          GenBank mRNA AJ278801.2 - 2 - 3 559 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004175.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lzts3

          Gene Ontology Categories:

          Function(s) Process(es)

          protein homooligomerization
          protein binding
          PDZ domain binding
          protein self-association

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P063485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946405 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P063483 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P063483 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P063483 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P572948 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10849932 - -
          Affymetrix Rat Gene 1.0 ST Array 10849931 - -
          Affymetrix Rat Gene 1.0 ST Array 10849929 - -
          Affymetrix Rat Gene 1.0 ST Array 10849930 - -
          Affymetrix Rat Gene 1.0 ST Array 10849928 - -
          Affymetrix Rat Gene 1.1 ST Array 10849927 - -
          Rat Genome 230 2.0 Array 1373057_PM_at - -
          Rat Genome 230 2.0 Array 1373057_at - -
          Affymetrix Rat Clariom S Assay PSR0300032441.rn.1 TC0300004175.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300032446.rn.1 TC0300004175.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300032442.rn.1 TC0300004175.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10849927 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21364576 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370419 - -
          RatRef-12 Expression BeadChip ILMN_60133 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline