Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00597121_m1
          See other VPS45 GE Assays ›
          Gene Symbol:
          Vps45
          Gene Name:
          vacuolar protein sorting 45 homolog (S. cerevisiae)
          Gene Aliases:
          Vsp45a
          Chromosome Location:
          Chr.2: 198123747 - 198184739 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00597121_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vps45
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172072.2 NP_742069.1 13 - 14 1548 64
          GenBank mRNA BC081705.1 - 13 - 14 1548 7192410 64
          U81160.1 - 13 - 14 1548 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004230.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vps45

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          vesicle docking involved in exocytosis
          biological_process
          protein transport
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P354806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137898 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145583 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92689 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81090 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95103 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23378 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135301 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30523 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P354806 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P354806 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P354806 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P354806 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700766184 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6196 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45306 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85715 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89141 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99485 - -
          Agilent Rat cDNA Microarray (G4105A) 700766184 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10825125 - -
          Affymetrix Rat Gene 1.0 ST Array 10825127 - -
          Affymetrix Rat Gene 1.0 ST Array 10825135 - -
          Affymetrix Rat Gene 1.0 ST Array 10825131 - -
          Affymetrix Rat Gene 1.0 ST Array 10825136 - -
          Affymetrix Rat Gene 1.0 ST Array 10825130 - -
          Affymetrix Rat Gene 1.0 ST Array 10825134 - -
          Affymetrix Rat Gene 1.0 ST Array 10825122 - -
          Affymetrix Rat Gene 1.0 ST Array 10825129 - -
          Affymetrix Rat Gene 1.0 ST Array 10825121 - -
          Affymetrix Rat Gene 1.0 ST Array 10825124 - -
          Affymetrix Rat Gene 1.0 ST Array 10825133 - -
          Affymetrix Rat Gene 1.0 ST Array 10825126 - -
          Affymetrix Rat Gene 1.0 ST Array 10825128 - -
          Affymetrix Rat Gene 1.0 ST Array 10825123 - -
          Affymetrix Rat Gene 1.0 ST Array 10825132 - -
          Affymetrix Rat Gene 1.1 ST Array 10825120 - -
          Rat Genome 230 2.0 Array 1370549_at - -
          Rat Genome 230 2.0 Array 1370549_PM_at - -
          Affymetrix HT Rat Focus Array 1370549_at - -
          Affymetrix HT Rat Focus Array 1370549_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200029095.rn.1 TC0200004230.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200029097.rn.1 TC0200004230.rn.1 RTA TC
          Rat Expression Set 230 1370549_PM_at - -
          Rat Expression Set 230 1370549_at - -
          Affymetrix Rat Gene 1.0 ST Array 10825120 - -
          Rat Genome U34 Set rc_AI227936_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22184052 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362991 - -
          RatRef-12 Expression BeadChip ILMN_52159 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline