Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00597135_m1
          See other TIMM10 GE Assays ›
          Gene Symbol:
          Timm10
          Gene Name:
          translocase of inner mitochondrial membrane 10 homolog (yeast)
          Gene Aliases:
          Tim10
          Chromosome Location:
          Chr.3: 72226566 - 72230087 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00597135_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Timm10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172074.3 NP_742071.1 1 - 2 637 85
          XM_006234473.2 XP_006234535.1 2 - 3 249 85
          XM_006234474.2 XP_006234536.1 3 - 4 361 85
          GenBank mRNA AF150091.1 - 1 - 2 72 85
          BC091116.1 - 1 - 2 638 7366212 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000975.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Timm10

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into mitochondrial inner membrane
          chaperone-mediated protein transport
          transporter activity
          protein transporter activity
          protein homodimerization activity
          metal ion binding
          chaperone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P561644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667201 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P561644 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P561644 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P561644 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P561644 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700480921 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20881 - -
          Agilent Rat cDNA Microarray (G4105A) 700480921 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10837349 - -
          Affymetrix Rat Gene 1.0 ST Array 10837350 - -
          Affymetrix Rat Gene 1.0 ST Array 10837348 - -
          Affymetrix Rat Gene 1.1 ST Array 10837347 - -
          Rat Genome 230 2.0 Array 1370366_PM_at - -
          Rat Genome 230 2.0 Array 1370366_at - -
          Affymetrix HT Rat Focus Array 1370366_PM_at - -
          Affymetrix HT Rat Focus Array 1370366_at - -
          Affymetrix Rat Clariom S Assay PSR0300007213.rn.1 TC0300000975.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300007215.rn.1 TC0300000975.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300007218.rn.1 TC0300000975.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300007214.rn.1 TC0300000975.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300007217.rn.1 TC0300000975.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300007212.rn.1 TC0300000975.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300007216.rn.1 TC0300000975.rn.1 RTA TC
          Rat Expression Set 230 1370366_at - -
          Rat Expression Set 230 1370366_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10837347 - -
          Rat Genome U34 Set rc_AI180367_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21474305 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361283 - -
          RatRef-12 Expression BeadChip ILMN_58834 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline