Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00597188_m1
          See other IMPA2 GE Assays ›
          Gene Symbol:
          Impa2
          Gene Name:
          inositol (myo)-1(or 4)-monophosphatase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 63016577 - 63050116 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00597188_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Impa2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172224.1 NP_757378.1 3 - 4 521 76
          XM_006254856.2 XP_006254918.1 3 - 4 3840 76
          GenBank mRNA AY160191.1 - 3 - 4 521 76
          BC083544.1 - 3 - 4 495 7191219 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000727.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Impa2

          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          inositol biosynthetic process
          biological_process
          phosphatidylinositol phosphorylation
          inositol phosphate dephosphorylation
          molecular_function
          inositol monophosphate 1-phosphatase activity
          protein homodimerization activity
          metal ion binding
          inositol monophosphate 3-phosphatase activity
          inositol monophosphate 4-phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P538334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425156 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42872 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47393 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50282 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124426 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P538334 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P538334 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P538334 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P538334 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64379 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66365 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47752 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10802409 - -
          Affymetrix Rat Gene 1.0 ST Array 10802412 - -
          Affymetrix Rat Gene 1.0 ST Array 10802413 - -
          Affymetrix Rat Gene 1.0 ST Array 10802410 - -
          Affymetrix Rat Gene 1.0 ST Array 10802411 - -
          Affymetrix Rat Gene 1.0 ST Array 10802415 - -
          Affymetrix Rat Gene 1.0 ST Array 10802414 - -
          Affymetrix Rat Gene 1.0 ST Array 10802408 - -
          Affymetrix Rat Gene 1.1 ST Array 10802407 - -
          Rat Genome 230 2.0 Array 1392467_PM_at - -
          Rat Genome 230 2.0 Array 1392467_at - -
          Affymetrix HT Rat Focus Array 1392467_PM_at - -
          Affymetrix HT Rat Focus Array 1392467_at - -
          Affymetrix Rat Clariom S Assay PSR1800005236.rn.1 TC1800000727.rn.1 RTA TC
          Rat Expression Set 230 1392467_at - -
          Rat Expression Set 230 1392467_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10802407 - -
          Rat Genome U34 Set rc_AA859981_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21177311 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367402 - -
          RatRef-12 Expression BeadChip ILMN_58534 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline