Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00597291_m1
          See other ZP4 GE Assays ›
          Gene Symbol:
          Zp4
          Gene Name:
          zona pellucida glycoprotein 4
          Gene Aliases:
          Zp-4
          Chromosome Location:
          Chr.17: 65497359 - 65503912 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00597291_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zp4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172330.3 NP_758833.2 6 - 7 891 98
          GenBank mRNA AF456325.1 - 6 - 7 892 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000869.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zp4

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of humoral immune response
          single fertilization
          protein kinase A signaling
          intracellular signal transduction
          positive regulation of T cell proliferation
          positive regulation of protein kinase activity
          acrosomal vesicle exocytosis
          protein kinase C signaling
          positive regulation of acrosome reaction
          negative regulation of binding of sperm to zona pellucida
          signal transducer activity
          acrosin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P032450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309985 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49835 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032450 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032450 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P032450 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P118635 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89225 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10799146 - -
          Affymetrix Rat Gene 1.0 ST Array 10799147 - -
          Affymetrix Rat Gene 1.0 ST Array 10799143 - -
          Affymetrix Rat Gene 1.0 ST Array 10799154 - -
          Affymetrix Rat Gene 1.0 ST Array 10799153 - -
          Affymetrix Rat Gene 1.0 ST Array 10799142 - -
          Affymetrix Rat Gene 1.0 ST Array 10799148 - -
          Affymetrix Rat Gene 1.0 ST Array 10799145 - -
          Affymetrix Rat Gene 1.0 ST Array 10799151 - -
          Affymetrix Rat Gene 1.0 ST Array 10799149 - -
          Affymetrix Rat Gene 1.0 ST Array 10799150 - -
          Affymetrix Rat Gene 1.0 ST Array 10799155 - -
          Affymetrix Rat Gene 1.0 ST Array 10799144 - -
          Affymetrix Rat Gene 1.0 ST Array 10799152 - -
          Affymetrix Rat Gene 1.1 ST Array 10799141 - -
          Affymetrix Rat Clariom S Assay PSR1700005887.rn.1 TC1700000869.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10799141 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21578094 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360086 - -
          RatRef-12 Expression BeadChip ILMN_67292 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline