Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00597295_m1
          See other CHMP3 GE Assays ›
          Gene Symbol:
          Chmp3
          Gene Name:
          charged multivesicular body protein 3
          Gene Aliases:
          Vps24
          Chromosome Location:
          Chr.4: 99449213 - 99496156 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00597295_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chmp3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172331.1 NP_758834.1 3 - 4 306 76
          XM_006236631.2 XP_006236693.1 3 - 4 415 76
          GenBank mRNA AY150169.1 - 3 - 4 306 76
          FQ215758.1 - 3 - 4 320 76
          FQ219218.1 - 3 - 4 325 76
          FQ234202.1 - 3 - 4 383 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001104.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chmp3

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell separation after cytokinesis
          nucleus organization
          mitotic metaphase plate congression
          endosome to lysosome transport
          regulation of centrosome duplication
          protein transport
          positive regulation of cytokinesis
          viral budding via host ESCRT complex
          regulation of viral process
          regulation of endosome size
          protein polymerization
          protein heterooligomerization
          cell division
          regulation of mitotic spindle assembly
          negative regulation of viral release from host cell
          positive regulation of viral release from host cell
          regulation of exosomal secretion
          phosphatidylinositol-4,5-bisphosphate binding
          phosphatidylcholine binding
          protein homodimerization activity
          ubiquitin-specific protease binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P285517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926957 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19669 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159077 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1680 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46048 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68911 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P285517 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P285517 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P285517 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P285517 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19322 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20868 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10634 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10856294 - -
          Affymetrix Rat Gene 1.0 ST Array 10856295 - -
          Affymetrix Rat Gene 1.0 ST Array 10856292 - -
          Affymetrix Rat Gene 1.0 ST Array 10856291 - -
          Affymetrix Rat Gene 1.0 ST Array 10856290 - -
          Affymetrix Rat Gene 1.0 ST Array 10856293 - -
          Affymetrix Rat Gene 1.1 ST Array 10856289 - -
          Rat Genome 230 2.0 Array 1389560_at - -
          Rat Genome 230 2.0 Array 1389560_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400007797.rn.1 TC0400001104.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10856289 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21841975 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375233 - -
          RatRef-12 Expression BeadChip ILMN_59677 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline