Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00597305_m1
          See other CTHRC1 GE Assays ›
          Gene Symbol:
          Cthrc1
          Gene Name:
          collagen triple helix repeat containing 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 77966722 - 77977004 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00597305_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cthrc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271300.1 NP_001258229.1 2 - 3 446 86
          NM_172333.2 NP_758836.1 2 - 3 491 86
          GenBank mRNA AY136824.1 - 2 - 3 493 86
          FQ220442.1 - 2 - 3 403 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001015.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cthrc1

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          cell migration
          positive regulation of protein binding
          positive regulation of osteoblast proliferation
          ossification involved in bone remodeling
          positive regulation of osteoblast differentiation
          Wnt signaling pathway, planar cell polarity pathway
          inner ear receptor stereocilium organization
          negative regulation of canonical Wnt signaling pathway
          cochlea morphogenesis
          establishment of planar polarity involved in neural tube closure
          frizzled binding
          Wnt-protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P144591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282430 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90915 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119753 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P144591 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P144591 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P144591 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P144591 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44367 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6457 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10896264 - -
          Affymetrix Rat Gene 1.0 ST Array 10896268 - -
          Affymetrix Rat Gene 1.0 ST Array 10896267 - -
          Affymetrix Rat Gene 1.0 ST Array 10896266 - -
          Affymetrix Rat Gene 1.1 ST Array 10896263 - -
          Rat Genome 230 2.0 Array 1384063_at - -
          Rat Genome 230 2.0 Array 1384063_PM_at - -
          Affymetrix HT Rat Focus Array 1384063_PM_at - -
          Affymetrix HT Rat Focus Array 1384063_at - -
          Affymetrix Rat Clariom S Assay PSR0700007626.rn.1 TC0700001015.rn.1 RTA TC
          Rat Expression Set 230 1384063_at - -
          Rat Expression Set 230 1384063_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10896263 - -
          Rat Genome U34 Set rc_AA958001_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21152303 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361478 - -
          RatRef-12 Expression BeadChip ILMN_66166 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline