Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00597360_m1
          See other MX1 GE Assays ›
          Gene Symbol:
          Mx1
          Gene Name:
          myxovirus (influenza virus) resistance 1
          Gene Aliases:
          IFI78
          Chromosome Location:
          Chr.11: 37891150 - 37916777 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00597360_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mx1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271058.1 NP_001257987.1 2 - 3 244 88
          NM_001271059.1 NP_001257988.1 3 - 4 288 88
          NM_001271060.1 NP_001257989.1 2 - 3 279 88
          NM_001271061.1 NP_001257990.1 2 - 3 213 88
          NM_001271062.1 NP_001257991.1 2 - 3 322 88
          NM_173096.3 NP_775119.2 2 - 3 402 88
          XM_006248146.2 XP_006248208.1 3 - 4 468 88
          XM_006248149.2 XP_006248211.1 3 - 4 511 88
          XM_008768555.1 XP_008766777.1 2 - 3 2404 88
          XM_008768556.1 XP_008766778.1 2 - 3 2377 88
          XM_008768557.1 XP_008766779.1 2 - 3 2312 88
          GenBank mRNA BC099784.1 - 2 - 3 390 7933587 88
          FQ226931.1 - 2 - 3 279 88
          FQ230542.1 - 2 - 3 291 88
          FQ231581.1 - 3 - 4 356 88
          X52711.1 - 1 - 2 196 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001396.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mx1

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          response to virus
          innate immune response
          defense response to virus
          GTPase activity
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P042415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950240 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70919 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35452 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042415 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042415 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P042415 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P328136 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700531064 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64771 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61923 - -
          Agilent Rat cDNA Microarray (G4105A) 700531064 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10753429 - -
          Affymetrix Rat Gene 1.0 ST Array 10753427 - -
          Affymetrix Rat Gene 1.0 ST Array 10753432 - -
          Affymetrix Rat Gene 1.0 ST Array 10753428 - -
          Affymetrix Rat Gene 1.0 ST Array 10753435 - -
          Affymetrix Rat Gene 1.0 ST Array 10753430 - -
          Affymetrix Rat Gene 1.0 ST Array 10753436 - -
          Affymetrix Rat Gene 1.0 ST Array 10753426 - -
          Affymetrix Rat Gene 1.0 ST Array 10753431 - -
          Affymetrix Rat Gene 1.0 ST Array 10753434 - -
          Affymetrix Rat Gene 1.0 ST Array 10753433 - -
          Affymetrix Rat Gene 1.1 ST Array 10753425 - -
          Rat Genome 230 2.0 Array 1371015_PM_at - -
          Rat Genome 230 2.0 Array 1371015_at - -
          Affymetrix HT Rat Focus Array 1371015_PM_at - -
          Affymetrix HT Rat Focus Array 1371015_at - -
          Affymetrix Rat Clariom S Assay PSR1100008457.rn.1 TC1100001396.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008455.rn.1 TC1100001396.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008464.rn.1 TC1100001396.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008466.rn.1 TC1100001396.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008458.rn.1 TC1100001396.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008459.rn.1 TC1100001396.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008456.rn.1 TC1100001396.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008454.rn.1 TC1100001396.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008467.rn.1 TC1100001396.rn.1 RTA TC
          Rat Expression Set 230 1371015_PM_at - -
          Rat Expression Set 230 1371015_at - -
          Affymetrix Rat Gene 1.0 ST Array 10753425 - -
          Rat Genome U34 Set X52711_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21684035 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373648 - -
          RatRef-12 Expression BeadChip ILMN_62633 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0