Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00597465_m1
          See other PARM1 GE Assays ›
          Gene Symbol:
          Parm1
          Gene Name:
          prostate androgen-regulated mucin-like protein 1
          Gene Aliases:
          CIPAR-1, Cipar1, PARM-1
          Chromosome Location:
          Chr.14: 17972914 - 18076870 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00597465_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Parm1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173114.1 NP_775137.1 2 - 3 905 66
          GenBank mRNA AJ010750.1 - 2 - 3 927 66
          BC060539.1 - 2 - 3 931 6890269 66
          FQ219991.1 - 2 - 3 928 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400001495.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Parm1

          Gene Ontology Categories:

          Function(s)

          apoptotic process
          positive regulation of telomerase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P505671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608350 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86040 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57949 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24852 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118131 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P505671 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P505671 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P505671 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1041365 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P505671 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701531153 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76268 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74531 - -
          Agilent Rat cDNA Microarray (G4105A) 701531153 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10775865 - -
          Affymetrix Rat Gene 1.0 ST Array 10775864 - -
          Affymetrix Rat Gene 1.0 ST Array 10775863 - -
          Affymetrix Rat Gene 1.1 ST Array 10775862 - -
          Rat Genome 230 2.0 Array 1372539_at - -
          Rat Genome 230 2.0 Array 1384985_at - -
          Rat Genome 230 2.0 Array 1384985_PM_at - -
          Rat Genome 230 2.0 Array 1370962_at - -
          Rat Genome 230 2.0 Array 1370962_PM_at - -
          Rat Genome 230 2.0 Array 1372539_PM_at - -
          Affymetrix HT Rat Focus Array 1370962_at - -
          Affymetrix HT Rat Focus Array 1370962_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400010993.rn.1 TC1400001495.rn.1 RTA TC
          Rat Expression Set 230 1370962_at - -
          Rat Expression Set 230 1370962_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10775862 - -
          Rat Genome U34 Set rc_AI136555_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20785651 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368730 - -
          RatRef-12 Expression BeadChip ILMN_56264 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline