Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00597574_m1
          See other CDK5RAP2 GE Assays ›
          Gene Symbol:
          Cdk5rap2
          Gene Name:
          CDK5 regulatory subunit associated protein 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 86387238 - 86554339 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00597574_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdk5rap2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173134.2 NP_775157.2 33 - 34 5248 68
          XM_006238273.2 XP_006238335.1 34 - 35 5733 68
          XM_006238274.2 XP_006238336.1 34 - 35 5727 68
          XM_006238276.2 XP_006238338.1 33 - 34 5635 68
          XM_006238277.2 XP_006238339.1 33 - 34 5629 68
          XM_006238278.2 XP_006238340.1 33 - 34 5482 68
          XM_006238279.2 XP_006238341.1 33 - 34 5475 68
          XM_006238280.2 XP_006238342.1 32 - 33 5404 68
          XM_008763760.1 XP_008761982.1 34 - 35 5733 68
          XM_008763761.1 XP_008761983.1 33 - 34 5602 68
          GenBank mRNA AF177478.1 - 2 - 3 286 68
          JX524852.1 - 34 - 35 5341 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002936.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdk5rap2

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of mitotic spindle orientation
          microtubule cytoskeleton organization
          microtubule bundle formation
          chromosome segregation
          brain development
          neurogenesis
          regulation of neuron differentiation
          negative regulation of neuron differentiation
          positive regulation of transcription, DNA-templated
          negative regulation of centriole replication
          centrosome organization
          calmodulin binding
          microtubule binding
          tubulin binding
          protein kinase binding
          protein complex binding
          transcription regulatory region DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P043309 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P024466 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P024466 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P043309 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P043309 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P024466 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P024466 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P043309 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P433349 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P615724 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P791077 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701438273 - -
          Agilent Rat cDNA Microarray (G4105A) 701438273 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10877589 - -
          Affymetrix Rat Gene 1.0 ST Array 10877591 - -
          Affymetrix Rat Gene 1.0 ST Array 10877586 - -
          Affymetrix Rat Gene 1.0 ST Array 10877588 - -
          Affymetrix Rat Gene 1.0 ST Array 10877592 - -
          Affymetrix Rat Gene 1.1 ST Array 10877585 - -
          Rat Genome 230 2.0 Array 1397832_PM_at - -
          Rat Genome 230 2.0 Array 1388419_at - -
          Rat Genome 230 2.0 Array 1388100_at - -
          Rat Genome 230 2.0 Array 1388100_PM_at - -
          Rat Genome 230 2.0 Array 1388419_PM_at - -
          Rat Genome 230 2.0 Array 1397832_at - -
          Affymetrix HT Rat Focus Array 1388100_at - -
          Affymetrix HT Rat Focus Array 1388100_PM_at - -
          Affymetrix HT Rat Focus Array 1388419_PM_at - -
          Affymetrix HT Rat Focus Array 1397832_at - -
          Affymetrix HT Rat Focus Array 1397832_PM_at - -
          Affymetrix HT Rat Focus Array 1388419_at - -
          Affymetrix Rat Clariom S Assay PSR0500021875.rn.1 TC0500002936.rn.1 RTA TC
          Rat Expression Set 230 1388100_PM_at - -
          Rat Expression Set 230 1388419_at - -
          Rat Expression Set 230 1388100_at - -
          Rat Expression Set 230 1388419_PM_at - -
          Rat Expression Set 230 1397832_PM_at - -
          Rat Expression Set 230 1397832_at - -
          Affymetrix Rat Gene 1.0 ST Array 10877585 - -
          Rat Genome U34 Set rc_AI171307_at - -
          Rat Genome U34 Set rc_AI235913_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21723808 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371600 - -
          RatRef-12 Expression BeadChip ILMN_57761 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline