Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00597609_m1
          See other DLGAP3 GE Assays ›
          Gene Symbol:
          Dlgap3
          Gene Name:
          discs, large (Drosophila) homolog-associated protein 3
          Gene Aliases:
          DAP-3, Dap3, SAPAP3
          Chromosome Location:
          Chr.5: 145237589 - 145304536 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00597609_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dlgap3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001276304.1 NP_001263233.1 2 - 3 1240 108
          XM_008763989.1 XP_008762211.1 3 - 4 2542 108
          GenBank mRNA AY530298.1 - 1 - 2 1103 108
          AY530299.1 - 1 - 2 1103 108
          U67139.1 - 2 - 3 1207 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001493.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein complex assembly
          cell-cell signaling
          beta-amyloid binding
          protein binding
          protein domain specific binding
          PDZ domain binding
          protein complex scaffold
          scaffold protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P072054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355052 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16123 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98542 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23478 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85645 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109365 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77648 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072054 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072054 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P072054 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P840999 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73281 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95728 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102936 - -
          Agilent Rat cDNA Microarray (G4105A) 701746912 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10872148 - -
          Affymetrix Rat Gene 1.0 ST Array 10872150 - -
          Affymetrix Rat Gene 1.0 ST Array 10872143 - -
          Affymetrix Rat Gene 1.0 ST Array 10872142 - -
          Affymetrix Rat Gene 1.0 ST Array 10872151 - -
          Affymetrix Rat Gene 1.0 ST Array 10872145 - -
          Affymetrix Rat Gene 1.0 ST Array 10872141 - -
          Affymetrix Rat Gene 1.0 ST Array 10872146 - -
          Affymetrix Rat Gene 1.0 ST Array 10872152 - -
          Affymetrix Rat Gene 1.0 ST Array 10872140 - -
          Affymetrix Rat Gene 1.0 ST Array 10872149 - -
          Affymetrix Rat Gene 1.0 ST Array 10872147 - -
          Affymetrix Rat Gene 1.0 ST Array 10872144 - -
          Affymetrix Rat Gene 1.1 ST Array 10872139 - -
          Rat Genome 230 2.0 Array 1370533_PM_at - -
          Rat Genome 230 2.0 Array 1370533_at - -
          Rat Genome 230 2.0 Array 1376233_at - -
          Rat Genome 230 2.0 Array 1376233_PM_at - -
          Affymetrix HT Rat Focus Array 1370533_PM_at - -
          Affymetrix HT Rat Focus Array 1376233_at - -
          Affymetrix HT Rat Focus Array 1376233_PM_at - -
          Affymetrix HT Rat Focus Array 1370533_at - -
          Affymetrix Rat Clariom S Assay PSR0500010574.rn.1 TC0500001493.rn.1 RTA TC
          Rat Expression Set 230 1370533_PM_at - -
          Rat Expression Set 230 1376233_at - -
          Rat Expression Set 230 1370533_at - -
          Rat Expression Set 230 1376233_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10872139 - -
          Rat Genome U34 Set U67139_at - -
          Rat Genome U34 Set rc_AA942850_s_at - -
          Rat Genome U34 Set rc_AI227989_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21160335 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369126 - -
          RatRef-12 Expression BeadChip ILMN_53184 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline