Search
Search
| Gene Symbol: Gimap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173153.2 | NP_775176.2 | 2 - 3 | 99 | 130 | |
| GenBank mRNA | AM285343.1 | - | 2 - 3 | 99 | 130 | |
| AM285683.1 | - | 2 - 3 | 99 | 130 | ||
| AY070268.1 | - | 1 - 2 | 81 | 130 | ||
| BC070952.1 | - | 2 - 3 | 87 | 7100695 | 130 | |
| CK479589.1 | - | 2 - 3 | 99 | 130 | ||
| DQ125339.1 | - | 2 - 3 | 88 | 130 | ||
| DQ125340.1 | - | 2 - 3 | 88 | 130 | ||
Set Membership: |
|