Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00597824_m1
          See other ABCA5 GE Assays ›
          Gene Symbol:
          Abca5
          Gene Name:
          ATP-binding cassette, subfamily A (ABC1), member 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 98573226 - 98645001 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00597824_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abca5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173307.1 NP_775429.1 17 - 18 2304 126
          XM_006247560.2 XP_006247622.1 17 - 18 2505 126
          GenBank mRNA AJ426052.2 - 17 - 18 2304 126
          AJ550165.1 - 16 - 17 2277 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000004041.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Abca5

          Panther Classification:

          Molecular Function -

          ATP-binding cassette (ABC) transporter primary active transporter transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          negative regulation of macrophage derived foam cell differentiation
          sterol transport
          cholesterol efflux
          high-density lipoprotein particle remodeling
          transporter activity
          ATP binding
          ATPase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P158609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854528 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60787 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92410 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60775 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48244 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67781 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54528 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42162 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61569 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160142 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113335 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P158609 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P158609 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P158609 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P158609 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72387 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49438 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31581 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80686 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82234 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10748741 - -
          Affymetrix Rat Gene 1.0 ST Array 10748732 - -
          Affymetrix Rat Gene 1.0 ST Array 10748739 - -
          Affymetrix Rat Gene 1.0 ST Array 10748729 - -
          Affymetrix Rat Gene 1.0 ST Array 10748722 - -
          Affymetrix Rat Gene 1.0 ST Array 10748738 - -
          Affymetrix Rat Gene 1.0 ST Array 10748735 - -
          Affymetrix Rat Gene 1.0 ST Array 10748731 - -
          Affymetrix Rat Gene 1.0 ST Array 10748740 - -
          Affymetrix Rat Gene 1.0 ST Array 10748725 - -
          Affymetrix Rat Gene 1.0 ST Array 10748726 - -
          Affymetrix Rat Gene 1.0 ST Array 10748742 - -
          Affymetrix Rat Gene 1.0 ST Array 10748752 - -
          Affymetrix Rat Gene 1.0 ST Array 10748723 - -
          Affymetrix Rat Gene 1.0 ST Array 10748736 - -
          Affymetrix Rat Gene 1.0 ST Array 10748751 - -
          Affymetrix Rat Gene 1.0 ST Array 10748744 - -
          Affymetrix Rat Gene 1.0 ST Array 10748719 - -
          Affymetrix Rat Gene 1.0 ST Array 10748730 - -
          Affymetrix Rat Gene 1.0 ST Array 10748755 - -
          Affymetrix Rat Gene 1.0 ST Array 10748721 - -
          Affymetrix Rat Gene 1.0 ST Array 10748728 - -
          Affymetrix Rat Gene 1.0 ST Array 10748749 - -
          Affymetrix Rat Gene 1.0 ST Array 10748743 - -
          Affymetrix Rat Gene 1.0 ST Array 10748720 - -
          Affymetrix Rat Gene 1.0 ST Array 10748724 - -
          Affymetrix Rat Gene 1.0 ST Array 10748750 - -
          Affymetrix Rat Gene 1.0 ST Array 10748727 - -
          Affymetrix Rat Gene 1.0 ST Array 10748753 - -
          Affymetrix Rat Gene 1.0 ST Array 10748734 - -
          Affymetrix Rat Gene 1.0 ST Array 10748756 - -
          Affymetrix Rat Gene 1.0 ST Array 10748748 - -
          Affymetrix Rat Gene 1.0 ST Array 10748746 - -
          Affymetrix Rat Gene 1.0 ST Array 10748754 - -
          Affymetrix Rat Gene 1.0 ST Array 10748733 - -
          Affymetrix Rat Gene 1.0 ST Array 10748745 - -
          Affymetrix Rat Gene 1.0 ST Array 10748747 - -
          Affymetrix Rat Gene 1.0 ST Array 10748737 - -
          Affymetrix Rat Gene 1.1 ST Array 10748718 - -
          Rat Genome 230 2.0 Array 1384469_at - -
          Rat Genome 230 2.0 Array 1384469_PM_at - -
          Affymetrix HT Rat Focus Array 1384469_PM_at - -
          Affymetrix HT Rat Focus Array 1384469_at - -
          Affymetrix Rat Clariom S Assay PSR1000036794.rn.1 TC1000004041.rn.1 RTA TC
          Rat Expression Set 230 1384469_at - -
          Rat Expression Set 230 1384469_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10748718 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20714794 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368155 - -
          RatRef-12 Expression BeadChip ILMN_49661 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline