Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00597907_m1
          See other TAOK1 GE Assays ›
          Gene Symbol:
          Taok1
          Gene Name:
          TAO kinase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 62542870 - 62630341 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00597907_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Taok1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173327.1 NP_775449.1 2 - 3 255 61
          GenBank mRNA AF084205.1 - 2 - 3 255 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001078.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Taok1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPKK activity
          DNA repair
          protein phosphorylation
          cellular response to DNA damage stimulus
          positive regulation of stress-activated MAPK cascade
          intracellular signal transduction
          positive regulation of protein kinase activity
          positive regulation of JNK cascade
          regulation of cytoskeleton organization
          execution phase of apoptosis
          protein serine/threonine kinase activity
          MAP kinase kinase kinase activity
          ATP binding
          protein kinase binding
          protein kinase activator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P191615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704189 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34789 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62299 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96248 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163786 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174469 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36603 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P191615 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P191615 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P191615 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P191615 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702211014 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3766 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6719 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35132 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17388 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34411 - -
          Agilent Rat cDNA Microarray (G4105A) 702211014 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10736429 - -
          Affymetrix Rat Gene 1.0 ST Array 10736431 - -
          Affymetrix Rat Gene 1.0 ST Array 10736430 - -
          Affymetrix Rat Gene 1.0 ST Array 10736426 - -
          Affymetrix Rat Gene 1.0 ST Array 10736424 - -
          Affymetrix Rat Gene 1.0 ST Array 10736428 - -
          Affymetrix Rat Gene 1.0 ST Array 10736437 - -
          Affymetrix Rat Gene 1.0 ST Array 10736439 - -
          Affymetrix Rat Gene 1.0 ST Array 10736438 - -
          Affymetrix Rat Gene 1.0 ST Array 10736423 - -
          Affymetrix Rat Gene 1.0 ST Array 10736434 - -
          Affymetrix Rat Gene 1.0 ST Array 10736436 - -
          Affymetrix Rat Gene 1.0 ST Array 10736427 - -
          Affymetrix Rat Gene 1.0 ST Array 10736432 - -
          Affymetrix Rat Gene 1.0 ST Array 10736435 - -
          Affymetrix Rat Gene 1.0 ST Array 10736422 - -
          Affymetrix Rat Gene 1.0 ST Array 10736421 - -
          Affymetrix Rat Gene 1.0 ST Array 10736433 - -
          Affymetrix Rat Gene 1.1 ST Array 10736420 - -
          Affymetrix Rat Gene 1.1 ST Array 10745378 - -
          Rat Genome 230 2.0 Array 1388002_PM_at - -
          Rat Genome 230 2.0 Array 1388002_at - -
          Affymetrix HT Rat Focus Array 1388002_PM_at - -
          Affymetrix HT Rat Focus Array 1388002_at - -
          Affymetrix Rat Clariom S Assay PSR1000009721.rn.1 TC1000001078.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009722.rn.1 TC1000001078.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009720.rn.1 TC1000001078.rn.1 RTA TC
          Rat Expression Set 230 1388002_at - -
          Rat Expression Set 230 1388002_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10736420 - -
          Rat Genome U34 Set AF084205_at - -
          Rat Genome U34 Set AF084205_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21431302 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354663 - -
          RatRef-12 Expression BeadChip ILMN_56731 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline