Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00597939_m1
          See other MAPK15 GE Assays ›
          Gene Symbol:
          Mapk15
          Gene Name:
          mitogen-activated protein kinase 15
          Gene Aliases:
          Erk7
          Chromosome Location:
          Chr.7: 117041358 - 117061796 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00597939_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mapk15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173331.2 NP_775453.1 1 - 2 110 84
          XM_006241777.2 XP_006241839.1 2 - 3 526 84
          XM_006241779.2 XP_006241841.1 2 - 3 526 84
          XM_008765547.1 XP_008763769.1 2 - 3 378 84
          GenBank mRNA AF078798.2 - 1 - 2 226 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001331.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mapk15

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          positive regulation of protein phosphorylation
          protein phosphorylation
          negative regulation of DNA replication
          positive regulation of protein ubiquitination
          response to estradiol
          positive regulation of protein catabolic process
          protein autophosphorylation
          MAP kinase activity
          protein binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P358227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683197 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P358227 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P358227 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P358227 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P358227 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12054 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88425 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10897125 - -
          Affymetrix Rat Gene 1.0 ST Array 10897138 - -
          Affymetrix Rat Gene 1.0 ST Array 10897134 - -
          Affymetrix Rat Gene 1.0 ST Array 10897126 - -
          Affymetrix Rat Gene 1.0 ST Array 10897131 - -
          Affymetrix Rat Gene 1.0 ST Array 10897130 - -
          Affymetrix Rat Gene 1.0 ST Array 10897137 - -
          Affymetrix Rat Gene 1.0 ST Array 10897127 - -
          Affymetrix Rat Gene 1.0 ST Array 10897140 - -
          Affymetrix Rat Gene 1.0 ST Array 10897136 - -
          Affymetrix Rat Gene 1.0 ST Array 10897132 - -
          Affymetrix Rat Gene 1.0 ST Array 10897128 - -
          Affymetrix Rat Gene 1.0 ST Array 10897139 - -
          Affymetrix Rat Gene 1.0 ST Array 10897135 - -
          Affymetrix Rat Gene 1.0 ST Array 10897133 - -
          Affymetrix Rat Gene 1.1 ST Array 10897121 - -
          Rat Genome 230 2.0 Array 1388062_at - -
          Rat Genome 230 2.0 Array 1388062_PM_at - -
          Affymetrix HT Rat Focus Array 1388062_PM_at - -
          Affymetrix HT Rat Focus Array 1388062_at - -
          Affymetrix Rat Clariom S Assay PSR0700009590.rn.1 TC0700001331.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700009589.rn.1 TC0700001331.rn.1 RTA TC
          Rat Expression Set 230 1388062_at - -
          Rat Expression Set 230 1388062_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10897121 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21858055 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349872 - -
          RatRef-12 Expression BeadChip ILMN_59674 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline