Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00598007_s1
          See other FZD5 GE Assays ›
          Gene Symbol:
          Fzd5
          Gene Name:
          frizzled class receptor 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 71443784 - 71445541 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00598007_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fzd5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173838.1 NP_776210.1 1 647 74
          GenBank mRNA AB052909.1 - 1 647 74

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Recommended Assay Rattus norvegicus Rat Wnt Panel 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fzd5

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          embryonic axis specification
          angiogenesis
          vasculature development
          G-protein coupled receptor signaling pathway
          multicellular organism development
          biological_process
          negative regulation of cell proliferation
          anterior/posterior axis specification, embryo
          Wnt signaling pathway
          embryonic camera-type eye development
          post-embryonic camera-type eye development
          positive regulation of interferon-gamma production
          T cell differentiation in thymus
          non-canonical Wnt signaling pathway
          positive regulation of JUN kinase activity
          Wnt signaling pathway involved in dorsal/ventral axis specification
          positive regulation of transcription from RNA polymerase II promoter
          cell maturation
          Spemann organizer formation
          canonical Wnt signaling pathway
          apoptotic process involved in morphogenesis
          branching involved in labyrinthine layer morphogenesis
          syncytiotrophoblast cell differentiation involved in labyrinthine layer development
          labyrinthine layer blood vessel development
          chorionic trophoblast cell differentiation
          regulation of canonical Wnt signaling pathway
          cellular response to molecule of bacterial origin
          regulation of chorionic trophoblast cell proliferation
          regulation of bicellular tight junction assembly
          G-protein coupled receptor activity
          Wnt-protein binding
          protein kinase binding
          ubiquitin protein ligase binding
          Wnt-activated receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P144257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626211 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P144257 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P144257 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P144257 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P144257 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10928534 - -
          Affymetrix Rat Gene 1.1 ST Array 10928533 - -
          Rat Genome 230 2.0 Array 1377796_PM_at - -
          Rat Genome 230 2.0 Array 1382360_PM_at - -
          Rat Genome 230 2.0 Array 1382360_at - -
          Rat Genome 230 2.0 Array 1377796_at - -
          Affymetrix HT Rat Focus Array 1382360_at - -
          Affymetrix HT Rat Focus Array 1382360_PM_at - -
          Affymetrix HT Rat Focus Array 1377796_PM_at - -
          Affymetrix HT Rat Focus Array 1377796_at - -
          Affymetrix Rat Clariom S Assay PSR0900017406.rn.1 TC0900002119.rn.1 RTA TC
          Rat Expression Set 230 1382360_at - -
          Rat Expression Set 230 1377796_at - -
          Rat Expression Set 230 1382360_PM_at - -
          Rat Expression Set 230 1377796_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10928533 - -
          Rat Genome U34 Set rc_AI180240_at - -
          Rat Genome U34 Set rc_AI234226_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20737838 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362768 - -
          RatRef-12 Expression BeadChip ILMN_48840 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline