Search
Search
| Gene Symbol: Cd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175577.3 | NP_783167.2 | 1 - 2 | 125 | 83 | |
| XM_006231021.2 | XP_006231083.1 | 1 - 2 | 232 | 83 | ||
| XM_008760218.1 | XP_008758440.1 | 1 - 2 | 232 | 83 | ||
| XM_008760219.1 | XP_008758441.1 | 1 - 2 | 231 | 83 | ||
| GenBank mRNA | AF488727.1 | - | 1 - 2 | 64 | 83 | |
| AY683561.1 | - | 1 - 2 | 47 | 83 | ||
| BC089767.1 | - | 1 - 2 | 125 | 7383438 | 83 | |
Set Membership: |
|