Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00598058_m1
          See other RCAN2 GE Assays ›
          Gene Symbol:
          Rcan2
          Gene Name:
          regulator of calcineurin 2
          Gene Aliases:
          Dcip2, Dscr1l1, Zaki-4
          Chromosome Location:
          Chr.9: 19518568 - 19749145 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00598058_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rcan2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175578.3 NP_783168.1 4 - 5 799 55
          XM_006244593.2 XP_006244655.1 3 - 4 723 55
          GenBank mRNA AF459022.1 - 3 - 4 521 55
          AF459023.1 - 4 - 5 641 55
          BC086974.1 - 4 - 5 799 7193921 55

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001651.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rcan2

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          response to oxidative stress
          short-term memory
          locomotion involved in locomotory behavior
          calcineurin-NFAT signaling cascade
          nucleotide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P084851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313997 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162486 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171757 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92580 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8765 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135243 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97383 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141637 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176488 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115483 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71470 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56396 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73401 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153496 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168114 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100803 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130479 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136158 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134017 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96646 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28407 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71651 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150213 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86581 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P325189 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P084851 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P084851 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P325189 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P325189 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P084851 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P325189 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P634930 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23390 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6083 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101450 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78362 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21426 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30979 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14603 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73884 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53574 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88384 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13758 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28440 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62930 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73825 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83211 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103479 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14369 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10926661 - -
          Affymetrix Rat Gene 1.0 ST Array 10926663 - -
          Affymetrix Rat Gene 1.0 ST Array 10926665 - -
          Affymetrix Rat Gene 1.0 ST Array 10926660 - -
          Affymetrix Rat Gene 1.0 ST Array 10926666 - -
          Affymetrix Rat Gene 1.0 ST Array 10926662 - -
          Affymetrix Rat Gene 1.0 ST Array 10926659 - -
          Affymetrix Rat Gene 1.0 ST Array 10926667 - -
          Affymetrix Rat Gene 1.1 ST Array 10926658 - -
          Rat Genome 230 2.0 Array 1374235_PM_at - -
          Rat Genome 230 2.0 Array 1374235_at - -
          Rat Genome 230 2.0 Array 1389066_at - -
          Rat Genome 230 2.0 Array 1389066_PM_at - -
          Affymetrix HT Rat Focus Array 1374235_at - -
          Affymetrix HT Rat Focus Array 1389066_PM_at - -
          Affymetrix HT Rat Focus Array 1389066_at - -
          Affymetrix HT Rat Focus Array 1374235_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900013562.rn.1 TC0900001651.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900013561.rn.1 TC0900001651.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900013563.rn.1 TC0900001651.rn.1 RTA TC
          Rat Expression Set 230 1389066_at - -
          Rat Expression Set 230 1374235_at - -
          Rat Expression Set 230 1374235_PM_at - -
          Rat Expression Set 230 1389066_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10926658 - -
          Rat Genome U34 Set rc_AI138048_at - -
          Rat Genome U34 Set rc_AI229304_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20722429 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375039 - -
          RatRef-12 Expression BeadChip ILMN_62637 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline