Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00598070_m1
          See other WNK4 GE Assays ›
          Gene Symbol:
          Wnk4
          Gene Name:
          WNK lysine deficient protein kinase 4
          Gene Aliases:
          Ac2-059, Prkwnk4
          Chromosome Location:
          Chr.10: 89181148 - 89198889 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00598070_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wnk4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175579.3 NP_783169.2 6 - 7 1492 75
          XM_006247278.2 XP_006247340.1 6 - 7 1597 75
          XM_006247279.2 XP_006247341.1 6 - 7 1597 75
          XM_006247280.2 XP_006247342.1 7 - 8 1004 75
          XM_006247281.2 XP_006247343.1 5 - 6 756 75
          GenBank mRNA AY187039.1 - 6 - 7 1469 75
          AY192567.1 - 6 - 7 1492 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000004367.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Wnk4

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          ion transport
          chloride transport
          protein localization
          intracellular signal transduction
          regulation of cellular process
          ion homeostasis
          renal sodium ion absorption
          distal tubule morphogenesis
          negative regulation of pancreatic juice secretion
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          chloride channel inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P155012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762889 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111588 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P155012 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P155012 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P155012 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P468909 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99356 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10738373 - -
          Affymetrix Rat Gene 1.0 ST Array 10738365 - -
          Affymetrix Rat Gene 1.0 ST Array 10738372 - -
          Affymetrix Rat Gene 1.0 ST Array 10738370 - -
          Affymetrix Rat Gene 1.0 ST Array 10738358 - -
          Affymetrix Rat Gene 1.0 ST Array 10738361 - -
          Affymetrix Rat Gene 1.0 ST Array 10738362 - -
          Affymetrix Rat Gene 1.0 ST Array 10738356 - -
          Affymetrix Rat Gene 1.0 ST Array 10738359 - -
          Affymetrix Rat Gene 1.0 ST Array 10738374 - -
          Affymetrix Rat Gene 1.0 ST Array 10738357 - -
          Affymetrix Rat Gene 1.0 ST Array 10738369 - -
          Affymetrix Rat Gene 1.0 ST Array 10738367 - -
          Affymetrix Rat Gene 1.0 ST Array 10738360 - -
          Affymetrix Rat Gene 1.0 ST Array 10738371 - -
          Affymetrix Rat Gene 1.0 ST Array 10738375 - -
          Affymetrix Rat Gene 1.0 ST Array 10738366 - -
          Affymetrix Rat Gene 1.0 ST Array 10738363 - -
          Affymetrix Rat Gene 1.0 ST Array 10738368 - -
          Affymetrix Rat Gene 1.0 ST Array 10738355 - -
          Affymetrix Rat Gene 1.1 ST Array 10738347 - -
          Rat Genome 230 2.0 Array 1371651_at - -
          Rat Genome 230 2.0 Array 1389662_PM_at - -
          Rat Genome 230 2.0 Array 1385834_PM_at - -
          Rat Genome 230 2.0 Array 1385834_at - -
          Rat Genome 230 2.0 Array 1371651_PM_at - -
          Rat Genome 230 2.0 Array 1389662_at - -
          Affymetrix HT Rat Focus Array 1385834_PM_at - -
          Affymetrix HT Rat Focus Array 1389662_PM_at - -
          Affymetrix HT Rat Focus Array 1385834_at - -
          Affymetrix HT Rat Focus Array 1389662_at - -
          Affymetrix Rat Clariom S Assay PSR1000040623.rn.1 TC1000004367.rn.1 RTA TC
          Rat Expression Set 230 1389662_at - -
          Rat Expression Set 230 1389662_PM_at - -
          Rat Expression Set 230 1385834_PM_at - -
          Rat Expression Set 230 1385834_at - -
          Affymetrix Rat Gene 1.0 ST Array 10738347 - -
          Rat Genome U34 Set rc_AI043880_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21585201 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361733 - -
          RatRef-12 Expression BeadChip ILMN_54686 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline