Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00598088_m1
          See other ID4 GE Assays ›
          Gene Symbol:
          Id4
          Gene Name:
          inhibitor of DNA binding 4
          Gene Aliases:
          Idb4
          Chromosome Location:
          Chr.17: 16692557 - 16695126 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00598088_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Id4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175582.1 NP_783172.1 2 - 3 499 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001463.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Id4

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          negative regulation of transcription from RNA polymerase II promoter
          neuroblast proliferation
          brain development
          circadian rhythm
          cell proliferation
          positive regulation of cell proliferation
          hippocampus development
          cerebral cortex neuron differentiation
          central nervous system myelination
          cellular protein localization
          fat cell differentiation
          negative regulation of fat cell differentiation
          negative regulation of neuron differentiation
          positive regulation of osteoblast differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of astrocyte differentiation
          negative regulation of oligodendrocyte differentiation
          RNA polymerase II transcription factor binding
          protein dimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P481915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728200 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P481915 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P481915 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P481915 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P481915 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P127869 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70700 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10797725 - -
          Affymetrix Rat Gene 1.0 ST Array 10797724 - -
          Affymetrix Rat Gene 1.0 ST Array 10797723 - -
          Affymetrix Rat Gene 1.1 ST Array 10797722 - -
          Rat Genome 230 2.0 Array 1375120_PM_at - -
          Rat Genome 230 2.0 Array 1375183_at - -
          Rat Genome 230 2.0 Array 1375120_at - -
          Rat Genome 230 2.0 Array 1375183_PM_at - -
          Rat Genome 230 2.0 Array 1381350_PM_at - -
          Rat Genome 230 2.0 Array 1385923_at - -
          Rat Genome 230 2.0 Array 1394022_PM_at - -
          Rat Genome 230 2.0 Array 1385923_PM_at - -
          Rat Genome 230 2.0 Array 1394022_at - -
          Rat Genome 230 2.0 Array 1381350_at - -
          Affymetrix HT Rat Focus Array 1375183_at - -
          Affymetrix HT Rat Focus Array 1375183_PM_at - -
          Affymetrix HT Rat Focus Array 1394022_PM_at - -
          Affymetrix HT Rat Focus Array 1375120_at - -
          Affymetrix HT Rat Focus Array 1385923_at - -
          Affymetrix HT Rat Focus Array 1385923_PM_at - -
          Affymetrix HT Rat Focus Array 1375120_PM_at - -
          Affymetrix HT Rat Focus Array 1394022_at - -
          Affymetrix Rat Clariom S Assay PSR1700010258.rn.1 TC1700001463.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700010259.rn.1 TC1700001463.rn.1 RTA TC
          Rat Expression Set 230 1375120_at - -
          Rat Expression Set 230 1375183_PM_at - -
          Rat Expression Set 230 1375183_at - -
          Rat Expression Set 230 1375120_PM_at - -
          Rat Expression Set 230 1394022_PM_at - -
          Rat Expression Set 230 1394022_at - -
          Rat Expression Set 230 1385923_at - -
          Rat Expression Set 230 1385923_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10797722 - -
          Rat Genome U34 Set rc_AI237207_at - -
          Rat Genome U34 Set rc_AA818860_at - -
          Rat Genome U34 Set rc_AI072481_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21591780 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline