Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00598233_m1
          See other IGF2BP1 GE Assays ›
          Gene Symbol:
          Igf2bp1
          Gene Name:
          insulin-like growth factor 2 mRNA binding protein 1
          Gene Aliases:
          Imp1, ZBP1
          Chromosome Location:
          Chr.10: 83774178 - 83823317 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00598233_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Igf2bp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175594.2 NP_783184.1 9 - 10 1301 71
          XM_006247193.2 XP_006247255.1 9 - 10 1622 71
          GenBank mRNA AF541940.1 - 9 - 10 1301 71
          BC161831.1 - 9 - 10 1333 9023972 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003645.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Igf2bp1

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA localization
          regulation of mRNA stability involved in response to stress
          negative regulation of translation
          pallium cell proliferation in forebrain
          mRNA transport
          CRD-mediated mRNA stabilization
          neuronal stem cell population maintenance
          nucleotide binding
          mRNA binding
          mRNA 3'-UTR binding
          poly(A) RNA binding
          translation regulator activity
          mRNA 5'-UTR binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P550215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244190 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179882 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167804 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20093 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44105 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6316 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P550215 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P550215 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P550215 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P550215 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58366 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14409 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58407 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90956 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10746565 - -
          Affymetrix Rat Gene 1.0 ST Array 10746557 - -
          Affymetrix Rat Gene 1.0 ST Array 10746566 - -
          Affymetrix Rat Gene 1.0 ST Array 10746568 - -
          Affymetrix Rat Gene 1.0 ST Array 10746569 - -
          Affymetrix Rat Gene 1.0 ST Array 10746564 - -
          Affymetrix Rat Gene 1.0 ST Array 10746571 - -
          Affymetrix Rat Gene 1.0 ST Array 10746563 - -
          Affymetrix Rat Gene 1.0 ST Array 10746562 - -
          Affymetrix Rat Gene 1.0 ST Array 10746560 - -
          Affymetrix Rat Gene 1.0 ST Array 10746561 - -
          Affymetrix Rat Gene 1.0 ST Array 10746567 - -
          Affymetrix Rat Gene 1.0 ST Array 10746558 - -
          Affymetrix Rat Gene 1.0 ST Array 10746559 - -
          Affymetrix Rat Gene 1.0 ST Array 10746570 - -
          Affymetrix Rat Gene 1.1 ST Array 10746556 - -
          Rat Genome 230 2.0 Array 1385469_PM_at - -
          Rat Genome 230 2.0 Array 1385469_at - -
          Affymetrix HT Rat Focus Array 1385469_PM_at - -
          Affymetrix HT Rat Focus Array 1385469_at - -
          Affymetrix Rat Clariom S Assay PSR1000032477.rn.1 TC1000003645.rn.1 RTA TC
          Rat Expression Set 230 1385469_at - -
          Rat Expression Set 230 1385469_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10746556 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21243503 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349651 - -
          RatRef-12 Expression BeadChip ILMN_68149 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline