Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00598283_m1
          See other SSX2IP GE Assays ›
          Gene Symbol:
          Ssx2ip
          Gene Name:
          synovial sarcoma, X breakpoint 2 interacting protein
          Gene Aliases:
          ADIP
          Chromosome Location:
          Chr.2: 252263358 - 252296140 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00598283_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ssx2ip
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175597.3 NP_783187.1 9 - 10 1291 83
          XM_006233454.2 XP_006233516.1 8 - 9 1104 83
          XM_006233455.2 XP_006233517.1 8 - 9 1253 83
          XM_006233458.2 XP_006233520.1 5 - 6 1234 83
          XM_008761498.1 XP_008759720.1 7 - 8 1159 83
          GenBank mRNA AF532970.1 - 7 - 8 1004 83
          BC078687.1 - 9 - 10 1291 7115887 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002371.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ssx2ip

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          actin cytoskeleton organization
          regulation of Rac protein signal transduction
          intraciliary transport involved in cilium morphogenesis
          cilium assembly
          centrosome organization
          regulation of cell motility
          protein domain specific binding
          protein binding, bridging

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P408679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397422 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175559 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56963 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88114 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P408679 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P408679 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P408679 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P995051 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P408679 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5894 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83863 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46242 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90522 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10819714 - -
          Affymetrix Rat Gene 1.0 ST Array 10819723 - -
          Affymetrix Rat Gene 1.0 ST Array 10819722 - -
          Affymetrix Rat Gene 1.0 ST Array 10819712 - -
          Affymetrix Rat Gene 1.0 ST Array 10819721 - -
          Affymetrix Rat Gene 1.0 ST Array 10819716 - -
          Affymetrix Rat Gene 1.0 ST Array 10819724 - -
          Affymetrix Rat Gene 1.0 ST Array 10819718 - -
          Affymetrix Rat Gene 1.0 ST Array 10819715 - -
          Affymetrix Rat Gene 1.0 ST Array 10819719 - -
          Affymetrix Rat Gene 1.0 ST Array 10819713 - -
          Affymetrix Rat Gene 1.0 ST Array 10819717 - -
          Affymetrix Rat Gene 1.0 ST Array 10819710 - -
          Affymetrix Rat Gene 1.0 ST Array 10819711 - -
          Affymetrix Rat Gene 1.0 ST Array 10819720 - -
          Affymetrix Rat Gene 1.1 ST Array 10819709 - -
          Rat Genome 230 2.0 Array 1373146_at - -
          Rat Genome 230 2.0 Array 1373146_PM_at - -
          Affymetrix HT Rat Focus Array 1373146_at - -
          Affymetrix HT Rat Focus Array 1373146_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200016656.rn.1 TC0200002371.rn.1 RTA TC
          Rat Expression Set 230 1373146_PM_at - -
          Rat Expression Set 230 1373146_at - -
          Affymetrix Rat Gene 1.0 ST Array 10819709 - -
          Rat Genome U34 Set rc_AI144877_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22256264 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349734 - -
          RatRef-12 Expression BeadChip ILMN_54244 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline