Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00598384_m1
          See other PPM1F GE Assays ›
          Gene Symbol:
          Ppm1f
          Gene Name:
          protein phosphatase, Mg2+/Mn2+ dependent, 1F
          Gene Aliases:
          CaMKPase
          Chromosome Location:
          Chr.11: 88313711 - 88343698 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00598384_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppm1f
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175755.2 NP_786931.1 5 - 6 829 94
          XM_006248674.1 XP_006248736.1 5 - 6 934 94
          XM_008768853.1 XP_008767075.1 5 - 6 880 94
          XM_008768854.1 XP_008767076.1 5 - 6 916 94
          XM_008768855.1 XP_008767077.1 5 - 6 934 94
          GenBank mRNA AB023634.1 - 5 - 6 829 94
          BC166854.1 - 5 - 6 934 9098491 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100002025.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppm1f

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein kinase activity
          protein dephosphorylation
          positive regulation of gene expression
          positive regulation of epithelial cell migration
          positive regulation of cell-substrate adhesion
          histone dephosphorylation
          negative regulation of peptidyl-serine phosphorylation
          cellular response to drug
          peptidyl-threonine dephosphorylation
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of protein kinase activity by regulation of protein phosphorylation
          negative regulation of transcription, DNA-templated
          positive regulation of growth
          positive regulation of chemotaxis
          positive regulation of stress fiber assembly
          positive regulation of focal adhesion assembly
          intrinsic apoptotic signaling pathway
          phosphoprotein phosphatase activity
          protein serine/threonine phosphatase activity
          phosphatase activity
          calmodulin-dependent protein phosphatase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P151675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112909 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100675 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10550 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105701 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P151675 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P151675 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P151675 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P123370 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81532 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78083 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35400 - -
          Agilent Rat cDNA Microarray (G4105A) 700510066 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10755815 - -
          Affymetrix Rat Gene 1.0 ST Array 10755819 - -
          Affymetrix Rat Gene 1.0 ST Array 10755816 - -
          Affymetrix Rat Gene 1.0 ST Array 10755818 - -
          Affymetrix Rat Gene 1.0 ST Array 10755820 - -
          Affymetrix Rat Gene 1.0 ST Array 10755821 - -
          Affymetrix Rat Gene 1.0 ST Array 10755817 - -
          Affymetrix Rat Gene 1.1 ST Array 10755814 - -
          Rat Genome 230 2.0 Array 1370552_at - -
          Rat Genome 230 2.0 Array 1370552_PM_at - -
          Rat Genome 230 2.0 Array 1398332_at - -
          Rat Genome 230 2.0 Array 1396266_at - -
          Rat Genome 230 2.0 Array 1396266_PM_at - -
          Rat Genome 230 2.0 Array 1398332_PM_at - -
          Affymetrix HT Rat Focus Array 1370552_PM_at - -
          Affymetrix HT Rat Focus Array 1398332_at - -
          Affymetrix HT Rat Focus Array 1396266_at - -
          Affymetrix HT Rat Focus Array 1396266_PM_at - -
          Affymetrix HT Rat Focus Array 1398332_PM_at - -
          Affymetrix HT Rat Focus Array 1370552_at - -
          Affymetrix Rat Clariom S Assay PSR1100013404.rn.1 TC1100002025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013407.rn.1 TC1100002025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013405.rn.1 TC1100002025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013403.rn.1 TC1100002025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013408.rn.1 TC1100002025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013410.rn.1 TC1100002025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013402.rn.1 TC1100002025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013406.rn.1 TC1100002025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013409.rn.1 TC1100002025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100013411.rn.1 TC1100002025.rn.1 RTA TC
          Rat Expression Set 230 1398332_at - -
          Rat Expression Set 230 1370552_PM_at - -
          Rat Expression Set 230 1398332_PM_at - -
          Rat Expression Set 230 1370552_at - -
          Rat Expression Set 230 1396266_at - -
          Rat Expression Set 230 1396266_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10755814 - -
          Rat Genome U34 Set rc_AA957669_at - -
          Rat Genome U34 Set rc_AI235644_at - -
          Rat Genome U34 Set rc_AI237137_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21518041 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371449 - -
          RatRef-12 Expression BeadChip ILMN_55902 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline