Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00598518_m1
          See other ABRA GE Assays ›
          Gene Symbol:
          Abra
          Gene Name:
          actin-binding Rho activating protein
          Gene Aliases:
          Ms1, Stars
          Chromosome Location:
          Chr.7: 80792615 - 80796683 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00598518_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abra
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175844.3 NP_787038.1 1 - 2 702 71
          GenBank mRNA AF336113.1 - 1 - 2 702 71
          BC158575.1 - 1 - 2 688 9033092 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003062.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Abra

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus, translocation
          transcription, DNA-templated
          positive regulation of Rho protein signal transduction
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of sequence-specific DNA binding transcription factor activity
          transcription coactivator activity
          actin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P600215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400008 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42345 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P600215 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P600215 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P600215 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P600215 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65428 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10903526 - -
          Affymetrix Rat Gene 1.0 ST Array 10903525 - -
          Affymetrix Rat Gene 1.1 ST Array 10903524 - -
          Rat Genome 230 2.0 Array 1390881_PM_at - -
          Rat Genome 230 2.0 Array 1390881_at - -
          Affymetrix HT Rat Focus Array 1390881_PM_at - -
          Affymetrix HT Rat Focus Array 1390881_at - -
          Affymetrix Rat Clariom S Assay PSR0700023642.rn.1 TC0700003062.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700023641.rn.1 TC0700003062.rn.1 RTA TC
          Rat Expression Set 230 1390881_PM_at - -
          Rat Expression Set 230 1390881_at - -
          Affymetrix Rat Gene 1.0 ST Array 10903524 - -
          Rat Genome U34 Set rc_AI172339_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22352862 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349223 - -
          RatRef-12 Expression BeadChip ILMN_68074 - -

          Back To Top
           

          Citations (1)

          Comparative in silico analysis identifies bona fide MyoD binding sites within the Myocyte stress 1 gene promoter.
          Authors
          Ounzain S, Dacwag CS, Samani NJ, Imbalzano AN, Chong NW
          Journal
          Bmc Mol Biol - Page No 50 (Tue Jan 01 00:00:00 EST 2008)
          Volume (Year)
          9 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline