Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00598544_m1
          See other C6 GE Assays ›
          Gene Symbol:
          C6
          Gene Name:
          complement component 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 54460329 - 54533801 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00598544_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_176074.3 NP_788263.1 3 - 4 444 109
          XM_008760747.1 XP_008758969.1 4 - 5 633 109
          XM_008760749.1 XP_008758971.1 4 - 5 618 109
          XM_008760750.1 XP_008758972.1 4 - 5 632 109
          GenBank mRNA AY230250.1 - 3 - 4 446 109

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_008760747.1, XM_008760749.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene C6

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          positive regulation of activation of membrane attack complex
          immune response
          complement activation, classical pathway
          cytolysis
          positive regulation of apoptotic process
          innate immune response
          positive regulation of angiogenesis
          positive regulation of complement activation
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185425 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84204 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17707 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94978 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26449 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116784 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105569 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117672 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170607 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14915 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14915 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14915 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14915 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4470 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79060 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40242 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87801 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30078 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10813267 - -
          Affymetrix Rat Gene 1.0 ST Array 10813263 - -
          Affymetrix Rat Gene 1.0 ST Array 10813260 - -
          Affymetrix Rat Gene 1.0 ST Array 10813266 - -
          Affymetrix Rat Gene 1.0 ST Array 10813270 - -
          Affymetrix Rat Gene 1.0 ST Array 10813257 - -
          Affymetrix Rat Gene 1.0 ST Array 10813258 - -
          Affymetrix Rat Gene 1.0 ST Array 10813269 - -
          Affymetrix Rat Gene 1.0 ST Array 10813268 - -
          Affymetrix Rat Gene 1.0 ST Array 10813256 - -
          Affymetrix Rat Gene 1.0 ST Array 10813254 - -
          Affymetrix Rat Gene 1.0 ST Array 10813265 - -
          Affymetrix Rat Gene 1.0 ST Array 10813262 - -
          Affymetrix Rat Gene 1.0 ST Array 10813255 - -
          Affymetrix Rat Gene 1.0 ST Array 10813261 - -
          Affymetrix Rat Gene 1.0 ST Array 10813264 - -
          Affymetrix Rat Gene 1.0 ST Array 10813259 - -
          Affymetrix Rat Gene 1.1 ST Array 10813253 - -
          Rat Genome 230 2.0 Array 1384580_PM_at - -
          Rat Genome 230 2.0 Array 1384580_at - -
          Affymetrix HT Rat Focus Array 1384580_PM_at - -
          Affymetrix HT Rat Focus Array 1384580_at - -
          Affymetrix Rat Clariom S Assay PSR0200002833.rn.1 TC0200000488.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200002829.rn.1 TC0200000488.rn.1 RTA TC
          Rat Expression Set 230 1384580_at - -
          Rat Expression Set 230 1384580_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10813253 - -
          Rat Genome U34 Set rc_AI045144_at - -
          Rat Genome U34 Set rc_AI045191_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21945938 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369281 - -
          RatRef-12 Expression BeadChip ILMN_68177 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline