Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00598568_m1
          See other CLIC6 GE Assays ›
          Gene Symbol:
          Clic6
          Gene Name:
          chloride intracellular channel 6
          Gene Aliases:
          Clic6b
          Chromosome Location:
          Chr.11: 32655653 - 32698004 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00598568_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clic6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_176078.2 NP_788267.1 3 - 4 1694 94
          GenBank mRNA AY191249.1 - 3 - 4 1694 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000285.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clic6

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of ion transmembrane transport
          chloride transmembrane transport
          voltage-gated ion channel activity
          chloride channel activity
          protein C-terminus binding
          D2 dopamine receptor binding
          D3 dopamine receptor binding
          D4 dopamine receptor binding
          protein homodimerization activity
          dopamine receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P449623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482295 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130441 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136708 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84666 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12882 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58181 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27208 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P449623 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P449623 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P449623 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P449623 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22099 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21823 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92887 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10750298 - -
          Affymetrix Rat Gene 1.0 ST Array 10750303 - -
          Affymetrix Rat Gene 1.0 ST Array 10750305 - -
          Affymetrix Rat Gene 1.0 ST Array 10750300 - -
          Affymetrix Rat Gene 1.0 ST Array 10750302 - -
          Affymetrix Rat Gene 1.0 ST Array 10750301 - -
          Affymetrix Rat Gene 1.0 ST Array 10750297 - -
          Affymetrix Rat Gene 1.0 ST Array 10750299 - -
          Affymetrix Rat Gene 1.0 ST Array 10750304 - -
          Affymetrix Rat Gene 1.1 ST Array 10750296 - -
          Affymetrix Rat Clariom S Assay PSR1100001243.rn.1 TC1100000285.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10750296 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21215032 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371422 - -
          RatRef-12 Expression BeadChip ILMN_50949 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline