Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00598583_m1
          See other SLC22A17 GE Assays ›
          Gene Symbol:
          Slc22a17
          Gene Name:
          solute carrier family 22, member 17
          Gene Aliases:
          24p3R, Boct, rBOCT
          Chromosome Location:
          Chr.15: 33569824 - 33576972 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00598583_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc22a17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177421.3 NP_803156.2 4 - 5 982 98
          XM_006251965.1 XP_006252027.1 4 - 5 1620 98
          GenBank mRNA AB040056.1 - 4 - 5 989 98
          BC099072.1 - 4 - 5 982 7312276 98
          FQ212273.1 - 4 - 5 984 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001686.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc22a17

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          ion transport
          signal transduction
          siderophore transport
          iron ion homeostasis
          transmembrane transport
          transmembrane signaling receptor activity
          transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P478500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419196 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45379 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P478500 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P478500 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P478500 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P478500 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16721 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10783673 - -
          Affymetrix Rat Gene 1.0 ST Array 10783670 - -
          Affymetrix Rat Gene 1.0 ST Array 10783675 - -
          Affymetrix Rat Gene 1.0 ST Array 10783674 - -
          Affymetrix Rat Gene 1.0 ST Array 10783677 - -
          Affymetrix Rat Gene 1.0 ST Array 10783678 - -
          Affymetrix Rat Gene 1.0 ST Array 10783676 - -
          Affymetrix Rat Gene 1.0 ST Array 10783680 - -
          Affymetrix Rat Gene 1.0 ST Array 10783679 - -
          Affymetrix Rat Gene 1.0 ST Array 10783671 - -
          Affymetrix Rat Gene 1.1 ST Array 10783669 - -
          Rat Genome 230 2.0 Array 1371889_at - -
          Rat Genome 230 2.0 Array 1371889_PM_at - -
          Affymetrix HT Rat Focus Array 1371889_at - -
          Affymetrix HT Rat Focus Array 1371889_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500010782.rn.1 TC1500001686.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010783.rn.1 TC1500001686.rn.1 RTA TC
          Rat Expression Set 230 1371889_at - -
          Rat Expression Set 230 1371889_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10783669 - -
          Rat Genome U34 Set rc_AA943588_at - -
          Rat Genome U34 Set rc_AA943588_g_at - -
          Rat Genome U34 Set rc_AI101977_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21434716 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371566 - -
          RatRef-12 Expression BeadChip ILMN_51112 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline