Search
Search
| Gene Symbol: Esrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107423.1 | NP_001100893.1 | 10 - 11 | 1532 | 85 | |
| XM_006255496.2 | XP_006255558.1 | 10 - 11 | 1402 | 85 | ||
| XM_008772499.1 | XP_008770721.1 | 9 - 10 | 1097 | 85 | ||
| GenBank mRNA | BC166804.1 | - | 10 - 11 | 1390 | 7938923 | 85 |
Set Membership: |
|