Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Axl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013147.1 | NP_001013165.1 | 13 - 14 | 1673 | 63 | |
NM_031794.1 | NP_113982.1 | 14 - 15 | 1700 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Recommended Assay Rattus norvegicus Inventoried Probe spans exons |