Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00665001_g1
          See other COX4I1 GE Assays ›
          Gene Symbol:
          Cox4i1
          Gene Name:
          cytochrome c oxidase subunit IV isoform 1
          Gene Aliases:
          Cox4, Cox4a
          Chromosome Location:
          Chr.19: 54245958 - 54252198 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00665001_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cox4i1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017202.1 NP_058898.1 4 - 5 457 72
          GenBank mRNA BC084719.1 - 3 - 4 373 7317602 72
          FQ211185.1 - 4 - 5 410 72
          FQ214265.1 - 4 - 5 466 72
          FQ217878.1 - 4 - 5 425 72
          FQ227128.1 - 4 - 5 455 72
          FQ228805.1 - 4 - 5 407 72
          X14209.1 - 4 - 5 439 72
          X15029.1 - 3 - 4 384 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000747.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cox4i1

          Panther Classification:

          Molecular Function -

          oxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to nutrient
          hydrogen ion transmembrane transport
          cytochrome-c oxidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P096569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881122 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P096569 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P096569 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P096569 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11856 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87888 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10808430 - -
          Affymetrix Rat Gene 1.0 ST Array 10808428 - -
          Affymetrix Rat Gene 1.0 ST Array 10808429 - -
          Affymetrix Rat Gene 1.0 ST Array 10808427 - -
          Affymetrix Rat Gene 1.1 ST Array 10808426 - -
          Rat Genome 230 2.0 Array 1367607_at - -
          Rat Genome 230 2.0 Array 1367607_PM_at - -
          Affymetrix HT Rat Focus Array 1367607_at - -
          Affymetrix HT Rat Focus Array 1367607_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900006150.rn.1 TC1900000747.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900006151.rn.1 TC1900000747.rn.1 RTA TC
          Rat Expression Set 230 1367607_at - -
          Rat Expression Set 230 1367607_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10808426 - -
          Rat Genome U34 Set rc_AA818226_s_at - -
          Rat Genome U34 Set J05425cds_s_at - -
          Rat Genome U34 Set X54081mRNA_i_at - -
          Rat Genome U34 Set X54081mRNA_i_at - -
          Rat Genome U34 Set rc_AA818226_s_at - -
          Rat Genome U34 Set J05425cds_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21858565 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372305 - -
          RatRef-12 Expression BeadChip ILMN_57807 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline