Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00667535_m1
          See other MSX1 GE Assays ›
          Gene Symbol:
          Msx1
          Gene Name:
          msh homeobox 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 77712262 - 77716061 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00667535_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Msx1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031059.1 NP_112321.1 1 - 2 593 83
          GenBank mRNA D83036.1 - 1 - 2 593 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000775.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Msx1

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cell morphogenesis
          in utero embryonic development
          epithelial to mesenchymal transition
          heart morphogenesis
          epithelial to mesenchymal transition involved in endocardial cushion formation
          transcription from RNA polymerase II promoter
          heart development
          muscle organ development
          negative regulation of cell proliferation
          anterior/posterior pattern specification
          mesenchymal cell proliferation
          pituitary gland development
          signal transduction involved in regulation of gene expression
          negative regulation of cell growth
          embryonic limb morphogenesis
          BMP signaling pathway
          positive regulation of BMP signaling pathway
          forebrain development
          midbrain development
          protein localization to nucleus
          embryonic forelimb morphogenesis
          embryonic hindlimb morphogenesis
          embryonic nail plate morphogenesis
          middle ear morphogenesis
          odontogenesis of dentin-containing tooth
          odontogenesis
          regulation of odontogenesis
          embryonic digit morphogenesis
          negative regulation of apoptotic process
          negative regulation of DNA binding
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          stem cell differentiation
          protein stabilization
          negative regulation of striated muscle cell differentiation
          cartilage development
          palate development
          face morphogenesis
          bone morphogenesis
          cartilage morphogenesis
          mammary gland epithelium development
          BMP signaling pathway involved in heart development
          cellular response to nicotine
          activation of meiosis
          positive regulation of intrinsic apoptotic signaling pathway by p53 class mediator
          negative regulation of transcription regulatory region DNA binding
          positive regulation of mesenchymal cell apoptotic process
          RNA polymerase II regulatory region sequence-specific DNA binding
          transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          p53 binding
          molecular_function
          DNA binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P571433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368208 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P571433 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P571433 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P571433 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P571433 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702023119 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43568 - -
          Agilent Rat cDNA Microarray (G4105A) 702023119 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10773254 - -
          Affymetrix Rat Gene 1.0 ST Array 10773253 - -
          Affymetrix Rat Gene 1.1 ST Array 10773252 - -
          Rat Genome 230 2.0 Array 1368302_PM_at - -
          Rat Genome 230 2.0 Array 1368302_at - -
          Affymetrix HT Rat Focus Array 1368302_PM_at - -
          Affymetrix HT Rat Focus Array 1368302_at - -
          Affymetrix Rat Clariom S Assay PSR1400005307.rn.1 TC1400000775.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400005306.rn.1 TC1400000775.rn.1 RTA TC
          Rat Expression Set 230 1368302_at - -
          Rat Expression Set 230 1368302_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10773252 - -
          Rat Genome U34 Set D83036_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22119128 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372537 - -
          RatRef-12 Expression BeadChip ILMN_51890 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline