Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00667825_m1
          See other VIM GE Assays ›
          Gene Symbol:
          Vim
          Gene Name:
          vimentin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 80882715 - 80891200 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00667825_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vim
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031140.1 NP_112402.1 8 - 9 1436 83
          GenBank mRNA BC061847.1 - 8 - 9 1476 5621065 83
          FQ220610.1 - 6 - 7 739 83
          FQ229240.1 - 6 - 7 743 83
          X62952.1 - 8 - 9 1436 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001048.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vim

          Gene Ontology Categories:

          Function(s) Process(es)

          aging
          response to radiation
          response to zinc ion
          response to selenium ion
          positive regulation of gene expression
          negative regulation of neuron projection development
          astrocyte development
          response to estradiol
          response to vitamin D
          skeletal muscle tissue regeneration
          cellular response to fibroblast growth factor stimulus
          intermediate filament-based process
          intermediate filament organization
          fat cell differentiation
          decidualization
          regulation of axonogenesis
          Bergmann glial cell differentiation
          positive regulation of glial cell proliferation
          SMAD protein signal transduction
          lens fiber cell development
          regulation of Schwann cell migration
          glycoprotein binding
          double-stranded RNA binding
          structural molecule activity
          structural constituent of cytoskeleton
          structural constituent of eye lens
          protein binding
          protein C-terminus binding
          kinase binding
          protein kinase binding
          identical protein binding
          protein phosphatase 2A binding
          scaffold protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P509365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912394 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145997 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P509365 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P509365 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P509365 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P509365 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700487041 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44719 - -
          Agilent Rat cDNA Microarray (G4105A) 700487041 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10796451 - -
          Affymetrix Rat Gene 1.0 ST Array 10796448 - -
          Affymetrix Rat Gene 1.0 ST Array 10796453 - -
          Affymetrix Rat Gene 1.0 ST Array 10796446 - -
          Affymetrix Rat Gene 1.0 ST Array 10796454 - -
          Affymetrix Rat Gene 1.0 ST Array 10796449 - -
          Affymetrix Rat Gene 1.0 ST Array 10796450 - -
          Affymetrix Rat Gene 1.0 ST Array 10796447 - -
          Affymetrix Rat Gene 1.0 ST Array 10796452 - -
          Affymetrix Rat Gene 1.1 ST Array 10796445 - -
          Rat Genome 230 2.0 Array 1367574_PM_at - -
          Rat Genome 230 2.0 Array 1367574_at - -
          Affymetrix HT Rat Focus Array 1367574_at - -
          Affymetrix HT Rat Focus Array 1367574_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700007183.rn.1 TC1700001048.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700007168.rn.1 TC1700001048.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700007175.rn.1 TC1700001048.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700007166.rn.1 TC1700001048.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700007179.rn.1 TC1700001048.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700007178.rn.1 TC1700001048.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700007167.rn.1 TC1700001048.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700007182.rn.1 TC1700001048.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700007169.rn.1 TC1700001048.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700007173.rn.1 TC1700001048.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700007172.rn.1 TC1700001048.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700007180.rn.1 TC1700001048.rn.1 RTA TC
          Rat Expression Set 230 1367574_PM_at - -
          Rat Expression Set 230 1367574_at - -
          Affymetrix Rat Gene 1.0 ST Array 10796445 - -
          Rat Genome U34 Set X62952_at - -
          Rat Genome U34 Set X62952_at - -
          Rat Genome U34 Set X62952_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21317299 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364377 - -
          RatRef-12 Expression BeadChip ILMN_54242 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline