Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00668206_g1
          See other OPRL1 GE Assays ›
          Gene Symbol:
          Oprl1
          Gene Name:
          opiate receptor-like 1
          Gene Aliases:
          KOR-3, KOR3, LC132, MOR-C, OFQR, ORL1, Oprl, XOR1
          Chromosome Location:
          Chr.3: 177225366 - 177231663 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00668206_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Oprl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031569.3 NP_113757.1 3 - 4 818 90
          XM_006235736.2 XP_006235798.1 4 - 5 908 90
          XM_006235738.2 XP_006235800.1 3 - 4 666 90
          XM_006235739.2 XP_006235801.1 5 - 6 942 90
          XM_008762474.1 XP_008760696.1 2 - 3 428 90
          GenBank mRNA AF115267.1 - 3 - 4 639 90
          AY152731.1 - 2 - 3 577 90
          D16438.1 - 3 - 4 629 90
          L28144.1 - 3 - 4 749 90
          L29419.1 - 2 - 3 712 90
          L33916.1 - 3 - 4 683 90
          U01913.1 - 3 - 4 758 90
          U05239.1 - 3 - 4 760 90
          U07871.1 - 2 - 3 608 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002610.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Oprl1

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          neuropeptide signaling pathway
          chemical synaptic transmission
          sensory perception of pain
          opioid receptor signaling pathway
          nociceptin receptor activity
          G-protein coupled receptor activity
          neuropeptide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P125150 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12591 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P408520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281384 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122294 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P125150 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P408520 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12591 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12591 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P408520 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P125150 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12591 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P408520 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P125150 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P408520 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12591 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103282 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10843095 - -
          Affymetrix Rat Gene 1.0 ST Array 10843094 - -
          Affymetrix Rat Gene 1.0 ST Array 10843091 - -
          Affymetrix Rat Gene 1.0 ST Array 10843093 - -
          Affymetrix Rat Gene 1.0 ST Array 10843088 - -
          Affymetrix Rat Gene 1.0 ST Array 10843086 - -
          Affymetrix Rat Gene 1.0 ST Array 10843099 - -
          Affymetrix Rat Gene 1.0 ST Array 10843096 - -
          Affymetrix Rat Gene 1.0 ST Array 10843097 - -
          Affymetrix Rat Gene 1.0 ST Array 10843087 - -
          Affymetrix Rat Gene 1.0 ST Array 10843098 - -
          Affymetrix Rat Gene 1.1 ST Array 10843084 - -
          Rat Genome 230 2.0 Array 1387661_a_at - -
          Rat Genome 230 2.0 Array 1369646_at - -
          Rat Genome 230 2.0 Array 1369645_at - -
          Rat Genome 230 2.0 Array 1369646_PM_at - -
          Rat Genome 230 2.0 Array 1387661_PM_a_at - -
          Rat Genome 230 2.0 Array 1369645_PM_at - -
          Affymetrix HT Rat Focus Array 1369645_at - -
          Affymetrix HT Rat Focus Array 1369646_at - -
          Affymetrix HT Rat Focus Array 1387661_a_at - -
          Affymetrix HT Rat Focus Array 1369646_PM_at - -
          Affymetrix HT Rat Focus Array 1387661_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369645_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300019373.rn.1 TC0300002610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300019377.rn.1 TC0300002610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300019375.rn.1 TC0300002610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300019369.rn.1 TC0300002610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300019370.rn.1 TC0300002610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300019374.rn.1 TC0300002610.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300019376.rn.1 TC0300002610.rn.1 RTA TC
          Rat Expression Set 230 1387661_PM_a_at - -
          Rat Expression Set 230 1369646_PM_at - -
          Rat Expression Set 230 1369645_PM_at - -
          Rat Expression Set 230 1369645_at - -
          Rat Expression Set 230 1369646_at - -
          Rat Expression Set 230 1387661_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10843084 - -
          Rat Genome U34 Set rc_AI229237_at - -
          Rat Genome U34 Set rc_AI229237_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21241066 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369970 - -
          RatRef-12 Expression BeadChip ILMN_47982 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline