Search
Search
| Gene Symbol: Igbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031624.3 | NP_113812.1 | 2 - 3 | 503 | 89 | |
| GenBank mRNA | AF000577.2 | - | 2 - 3 | 495 | 89 | |
| BC068202.1 | - | 2 - 3 | 507 | 6921265 | 89 | |
| FQ212697.1 | - | 2 - 3 | 512 | 89 | ||
| FQ222671.1 | - | 2 - 3 | 505 | 89 | ||
Set Membership: |
|