Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00670241_m1
          See other NUP210 GE Assays ›
          Gene Symbol:
          Nup210
          Gene Name:
          nucleoporin 210
          Gene Aliases:
          Pom210
          Chromosome Location:
          Chr.4: 122643952 - 122741198 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00670241_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nup210
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053322.2 NP_445774.1 23 - 24 3255 60
          XM_006236921.2 XP_006236983.1 21 - 22 3436 60
          GenBank mRNA Y00826.1 - 23 - 24 3256 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003332.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nup210

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          protein transport
          mRNA transport
          structural constituent of nuclear pore
          protein dimerization activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P512828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936837 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36112 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74323 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90926 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52122 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136537 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112648 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4167 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66779 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114172 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69340 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87205 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173505 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P512828 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P512828 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P512828 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P512828 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702150236 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70334 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14672 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77171 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69982 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86323 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46107 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21317 - -
          Agilent Rat cDNA Microarray (G4105A) 702150236 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10864064 - -
          Affymetrix Rat Gene 1.0 ST Array 10864049 - -
          Affymetrix Rat Gene 1.0 ST Array 10864067 - -
          Affymetrix Rat Gene 1.0 ST Array 10864063 - -
          Affymetrix Rat Gene 1.0 ST Array 10864076 - -
          Affymetrix Rat Gene 1.0 ST Array 10864075 - -
          Affymetrix Rat Gene 1.0 ST Array 10864091 - -
          Affymetrix Rat Gene 1.0 ST Array 10864054 - -
          Affymetrix Rat Gene 1.0 ST Array 10864060 - -
          Affymetrix Rat Gene 1.0 ST Array 10864087 - -
          Affymetrix Rat Gene 1.0 ST Array 10864086 - -
          Affymetrix Rat Gene 1.0 ST Array 10864066 - -
          Affymetrix Rat Gene 1.0 ST Array 10864057 - -
          Affymetrix Rat Gene 1.0 ST Array 10864058 - -
          Affymetrix Rat Gene 1.0 ST Array 10864078 - -
          Affymetrix Rat Gene 1.0 ST Array 10864085 - -
          Affymetrix Rat Gene 1.0 ST Array 10864053 - -
          Affymetrix Rat Gene 1.0 ST Array 10864065 - -
          Affymetrix Rat Gene 1.0 ST Array 10864082 - -
          Affymetrix Rat Gene 1.0 ST Array 10864081 - -
          Affymetrix Rat Gene 1.0 ST Array 10864090 - -
          Affymetrix Rat Gene 1.0 ST Array 10864061 - -
          Affymetrix Rat Gene 1.0 ST Array 10864074 - -
          Affymetrix Rat Gene 1.0 ST Array 10864071 - -
          Affymetrix Rat Gene 1.0 ST Array 10864051 - -
          Affymetrix Rat Gene 1.0 ST Array 10864083 - -
          Affymetrix Rat Gene 1.0 ST Array 10864055 - -
          Affymetrix Rat Gene 1.0 ST Array 10864079 - -
          Affymetrix Rat Gene 1.0 ST Array 10864070 - -
          Affymetrix Rat Gene 1.0 ST Array 10864062 - -
          Affymetrix Rat Gene 1.0 ST Array 10864072 - -
          Affymetrix Rat Gene 1.0 ST Array 10864050 - -
          Affymetrix Rat Gene 1.0 ST Array 10864059 - -
          Affymetrix Rat Gene 1.0 ST Array 10864089 - -
          Affymetrix Rat Gene 1.0 ST Array 10864068 - -
          Affymetrix Rat Gene 1.0 ST Array 10864077 - -
          Affymetrix Rat Gene 1.0 ST Array 10864073 - -
          Affymetrix Rat Gene 1.0 ST Array 10864080 - -
          Affymetrix Rat Gene 1.0 ST Array 10864088 - -
          Affymetrix Rat Gene 1.0 ST Array 10864052 - -
          Affymetrix Rat Gene 1.0 ST Array 10864056 - -
          Affymetrix Rat Gene 1.1 ST Array 10864048 - -
          Rat Genome 230 2.0 Array 1367919_PM_at - -
          Rat Genome 230 2.0 Array 1384596_at - -
          Rat Genome 230 2.0 Array 1384596_PM_at - -
          Rat Genome 230 2.0 Array 1367919_at - -
          Affymetrix HT Rat Focus Array 1367919_at - -
          Affymetrix HT Rat Focus Array 1367919_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400023675.rn.1 TC0400003332.rn.1 RTA TC
          Rat Expression Set 230 1367919_PM_at - -
          Rat Expression Set 230 1367919_at - -
          Affymetrix Rat Gene 1.0 ST Array 10864048 - -
          Rat Genome U34 Set Y00826_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370368 - -
          RatRef-12 Expression BeadChip ILMN_57726 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline